Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445474_m1
          See other AK1 GE Assays ›
          Gene Symbol:
          Ak1
          Gene Name:
          adenylate kinase 1
          Gene Aliases:
          Ak-1, B430205N08Rik
          Chromosome Location:
          Chr.2: 32621758 - 32635058 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445474_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ak1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001198790.1 NP_001185719.1 4 - 5 360 107
          NM_001198791.1 NP_001185720.1 4 - 5 342 107
          NM_001198792.1 NP_001185721.1 3 - 4 301 107
          NM_021515.3 NP_067490.1 3 - 4 319 107
          GenBank mRNA AJ010108.1 - 3 - 4 225 107
          AJ010109.1 - 3 - 4 315 107
          AK046613.1 - 4 - 5 361 107
          AK089270.1 - 4 - 5 343 107
          BC014802.1 - 4 - 5 286 4505142 107
          BC054366.1 - 3 - 4 308 5346232 107
          CF950379.1 - 3 - 4 319 107
          DQ486026.1 - 4 - 5 373 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005471.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ak1

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleobase-containing compound metabolic process
          nucleoside diphosphate phosphorylation
          ADP biosynthetic process
          nucleoside triphosphate biosynthetic process
          positive regulation of glucose transmembrane transport
          phosphorylation
          AMP metabolic process
          ATP metabolic process
          inosine biosynthetic process
          nucleotide phosphorylation
          nucleotide binding
          adenylate kinase activity
          nucleoside diphosphate kinase activity
          ATP binding
          kinase activity
          transferase activity
          phosphotransferase activity, phosphate group as acceptor
          nucleobase-containing compound kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200005367.mm.1 TC0200005471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005367.mm.1 NM_001198791.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005367.mm.1 NM_001198791.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005370.mm.1 TC0200005471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005370.mm.1 NM_001198790.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005370.mm.1 NM_001198790.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005370.mm.1 NM_001198791.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005370.mm.1 NM_001198791.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005370.mm.1 NM_001198792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005370.mm.1 NM_001198792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005368.mm.1 TC0200005471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005368.mm.1 NM_001198791.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005368.mm.1 NM_001198791.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005376.mm.1 TC0200005471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005376.mm.1 NM_001198790.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005376.mm.1 NM_001198790.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005376.mm.1 NM_001198791.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005376.mm.1 NM_001198791.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005376.mm.1 NM_001198792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005376.mm.1 NM_001198792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005376.mm.1 NM_021515.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005376.mm.1 NM_021515.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005378.mm.1 TC0200005471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005378.mm.1 NM_001198790.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005378.mm.1 NM_001198790.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005378.mm.1 NM_001198791.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005378.mm.1 NM_001198791.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005378.mm.1 NM_001198792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005378.mm.1 NM_001198792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005378.mm.1 NM_021515.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005378.mm.1 NM_021515.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005366.mm.1 TC0200005471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005366.mm.1 NM_001198790.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005366.mm.1 NM_001198790.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005369.mm.1 TC0200005471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005369.mm.1 NM_001198792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005369.mm.1 NM_001198792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005374.mm.1 TC0200005471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005374.mm.1 NM_021515.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005374.mm.1 NM_021515.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005377.mm.1 TC0200005471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005377.mm.1 NM_001198790.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005377.mm.1 NM_001198790.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005377.mm.1 NM_001198791.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005377.mm.1 NM_001198791.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005377.mm.1 NM_001198792.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005377.mm.1 NM_001198792.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005377.mm.1 NM_021515.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005377.mm.1 NM_021515.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0