Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00445770_m1
          See other COL4A3BP GE Assays ›
          Gene Symbol:
          Col4a3bp
          Gene Name:
          collagen, type IV, alpha 3 (Goodpasture antigen) binding protein
          Gene Aliases:
          2810404O15Rik, 9230101K08Rik, AU016711, CERT, GPBP
          Chromosome Location:
          Chr.13: 96542735 - 96640167 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00445770_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Col4a3bp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164222.1 NP_001157694.1 16 - 17 2240 137
          NM_023420.2 NP_075909.1 17 - 18 2318 137
          GenBank mRNA AF232932.1 - 17 - 18 2332 137
          AF232934.1 - 16 - 17 2254 137
          AK012989.1 - 16 - 17 2221 137
          AK018103.1 - 5 - 6 609 137
          AK030305.1 - 17 - 18 2148 137

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001089.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Col4a3bp

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          in utero embryonic development
          heart morphogenesis
          ceramide metabolic process
          transport
          lipid transport
          muscle contraction
          endoplasmic reticulum organization
          signal transduction
          cell population proliferation
          phosphorylation
          response to endoplasmic reticulum stress
          ER to Golgi ceramide transport
          lipid homeostasis
          mitochondrion morphogenesis
          lipid binding
          kinase activity
          phosphatidylinositol-4-phosphate binding
          ceramide binding
          ceramide transfer activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300006943.mm.1 TC1300001089.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300006943.mm.1 NM_001164222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006943.mm.1 NM_001164222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006943.mm.1 NM_023420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006943.mm.1 NM_023420.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006944.mm.1 TC1300001089.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300006944.mm.1 NM_001164222.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006944.mm.1 NM_001164222.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006944.mm.1 NM_023420.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300006944.mm.1 NM_023420.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0