Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026508.2 | NP_080784.1 | 1 - 2 | 117 | 89 | |
GenBank mRNA | AK002409.1 | - | 1 - 2 | 103 | 89 | |
AK010341.1 | - | 1 - 2 | 117 | 89 | ||
AK088471.1 | - | 1 - 2 | 106 | 89 | ||
AK143494.1 | - | 1 - 2 | 135 | 89 | ||
AK162067.1 | - | 1 - 2 | 105 | 89 | ||
AK167181.1 | - | 1 - 2 | 106 | 89 | ||
BC006685.1 | - | 1 - 2 | 85 | 89 | ||
BC006864.1 | - | 1 - 2 | 85 | 89 | ||
BC022912.1 | - | 1 - 2 | 114 | 4242394 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |