Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026508.2 | NP_080784.1 | 7 - 8 | 848 | 88 | |
GenBank mRNA | AK002409.1 | - | 7 - 8 | 834 | 88 | |
AK010341.1 | - | 7 - 8 | 848 | 88 | ||
AK088471.1 | - | 7 - 8 | 837 | 88 | ||
AK143494.1 | - | 7 - 8 | 866 | 88 | ||
AK162067.1 | - | 7 - 8 | 836 | 88 | ||
AK167181.1 | - | 7 - 8 | 837 | 88 | ||
BC006685.1 | - | 7 - 8 | 816 | 88 | ||
BC006864.1 | - | 7 - 8 | 816 | 88 | ||
BC022912.1 | - | 7 - 8 | 845 | 4242394 | 88 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |