Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290389.1 | NP_001277318.1 | 1 - 2 | 212 | 85 | |
NM_133690.3 | NP_598451.1 | 1 - 2 | 212 | 85 | ||
GenBank mRNA | AF348324.1 | - | 1 - 2 | 67 | 85 | |
AF348325.1 | - | 1 - 2 | 67 | 85 | ||
AK053500.1 | - | 1 - 2 | 151 | 85 | ||
BC137923.1 | - | 1 - 2 | 165 | 8860942 | 85 | |
BC137924.1 | - | 1 - 2 | 165 | 8860943 | 85 | |
BY771893.1 | - | 1 - 2 | 212 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |