Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00447759_mH
          See other EZR GE Assays ›
          Gene Symbol:
          Ezr
          Gene Name:
          ezrin
          Gene Aliases:
          AW146364, R75297, Vil2, p81
          Chromosome Location:
          Chr.17: 6738131 - 6782780 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00447759_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ezr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009510.2 NP_033536.2 9 - 10 1232 66
          GenBank mRNA AK002766.1 - 9 - 10 1206 66
          AK035271.1 - 5 - 6 600 66
          AK135194.1 - 9 - 10 1208 66
          AK135353.1 - 9 - 10 1208 66
          AK145707.1 - 9 - 10 1233 66
          AK159717.1 - 9 - 10 1210 66
          AK159824.1 - 9 - 10 1208 66
          AK159937.1 - 10 - 11 1184 66
          AK168410.1 - 9 - 10 1211 66
          AK168840.1 - 9 - 10 1208 66
          AK170610.1 - 9 - 10 1208 66
          AK172336.1 - 9 - 10 1215 66
          BC048181.1 - 9 - 10 1195 6826190 66
          BC098502.1 - 9 - 10 1184 30047137 66
          X60671.1 - 9 - 10 1190 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001390.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ezr

          Gene Ontology Categories:

          Function(s) Process(es)

          intestinal D-glucose absorption
          sphingosine-1-phosphate receptor signaling pathway
          regulation of cell shape
          positive regulation of gene expression
          microvillus assembly
          actin cytoskeleton reorganization
          receptor internalization
          regulation of microvillus length
          establishment or maintenance of apical/basal cell polarity
          positive regulation of multicellular organism growth
          filopodium assembly
          phosphatidylinositol-mediated signaling
          positive regulation of protein secretion
          actin filament bundle assembly
          establishment of endothelial barrier
          regulation of organelle assembly
          terminal web assembly
          positive regulation of protein localization to early endosome
          positive regulation of protein localization to plasma membrane
          positive regulation of cellular protein catabolic process
          positive regulation of early endosome to late endosome transport
          RNA binding
          actin binding
          protein binding
          cytoskeletal protein binding
          protein domain specific binding
          protein-containing complex binding
          cell adhesion molecule binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700012541.mm.1 TC1700001390.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012541.mm.1 NM_009510.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012541.mm.1 NM_009510.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012542.mm.1 TC1700001390.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700012542.mm.1 NM_009510.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700012542.mm.1 NM_009510.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0