Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159717.1 | NP_001153189.1 | 3 - 4 | 228 | 122 | |
NM_001159718.1 | NP_001153190.1 | 2 - 3 | 201 | 122 | ||
NM_001159719.1 | NP_001153191.1 | 3 - 4 | 276 | 122 | ||
NM_010891.2 | NP_035021.1 | 3 - 4 | 232 | 122 | ||
GenBank mRNA | AK028072.1 | - | 3 - 4 | 214 | 122 | |
AK146616.1 | - | 3 - 4 | 229 | 122 | ||
AK151591.1 | - | 3 - 4 | 216 | 122 | ||
AK169852.1 | - | 2 - 3 | 251 | 122 | ||
AK171331.1 | - | 3 - 4 | 211 | 122 | ||
BB849650.1 | - | 3 - 4 | 202 | 122 | ||
BC138636.1 | - | 2 - 3 | 169 | 8861656 | 122 | |
BC138637.1 | - | 2 - 3 | 169 | 8861657 | 122 | |
CJ075082.1 | - | 3 - 4 | 255 | 122 | ||
D49382.2 | - | 3 - 4 | 187 | 122 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |