Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00448948_m1
          See other TBX1 GE Assays ›
          Gene Symbol:
          Tbx1
          Gene Name:
          T-box 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 18581704 - 18590671 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00448948_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001285472.1 NP_001272401.1 3 - 4 505 88
          NM_011532.2 NP_035662.1 1 - 2 433 88
          GenBank mRNA AF326960.2 - 1 - 2 433 88
          AF349658.1 - 3 - 4 505 88
          BC139474.1 - 1 - 2 433 8862496 88
          U57327.1 - 1 - 2 55 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001317.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tbx1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          angiogenesis
          blood vessel development
          branching involved in blood vessel morphogenesis
          cell fate specification
          neural crest cell migration
          positive regulation of protein phosphorylation
          lymph vessel development
          blood vessel remodeling
          positive regulation of mesenchymal cell proliferation
          heart morphogenesis
          outflow tract septum morphogenesis
          outflow tract morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          multicellular organism development
          determination of left/right symmetry
          pattern specification process
          mesoderm development
          heart development
          muscle organ development
          sensory perception of sound
          cell population proliferation
          positive regulation of cell population proliferation
          anterior/posterior pattern specification
          vagus nerve morphogenesis
          epithelial cell differentiation
          thyroid gland development
          social behavior
          aorta morphogenesis
          ear morphogenesis
          inner ear morphogenesis
          outer ear morphogenesis
          middle ear morphogenesis
          odontogenesis of dentin-containing tooth
          muscle cell fate commitment
          positive regulation of MAPK cascade
          tongue morphogenesis
          cellular response to fibroblast growth factor stimulus
          negative regulation of cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          retinoic acid receptor signaling pathway
          blood vessel morphogenesis
          thymus development
          muscle organ morphogenesis
          embryonic cranial skeleton morphogenesis
          embryonic viscerocranium morphogenesis
          semicircular canal morphogenesis
          artery morphogenesis
          positive regulation of epithelial cell proliferation
          parathyroid gland development
          soft palate development
          pharyngeal system development
          face morphogenesis
          muscle tissue morphogenesis
          coronary artery morphogenesis
          enamel mineralization
          otic vesicle morphogenesis
          positive regulation of secondary heart field cardioblast proliferation
          cochlea morphogenesis
          mesenchymal cell apoptotic process
          regulation of animal organ morphogenesis
          positive regulation of tongue muscle cell differentiation
          negative regulation of mesenchymal cell apoptotic process
          RNA polymerase II intronic transcription regulatory region sequence-specific DNA binding
          DNA-binding transcription activator activity, RNA polymerase II-specific
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          protein homodimerization activity
          sequence-specific DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600010866.mm.1 TC1600001317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010866.mm.1 NM_001285476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010866.mm.1 NM_011532.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010863.mm.1 TC1600001317.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010863.mm.1 NM_001285472.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010863.mm.1 NM_001285476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010863.mm.1 NM_011532.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix