Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100020931.mm.1 |
TC1100002154.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020931.mm.1 |
NM_017401.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020931.mm.1 |
NM_017401.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020935.mm.1 |
TC1100002154.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020935.mm.1 |
NM_017401.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020935.mm.1 |
NM_017401.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020927.mm.1 |
TC1100002154.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020927.mm.1 |
NM_017401.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020927.mm.1 |
NM_017401.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020932.mm.1 |
TC1100002154.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020932.mm.1 |
NM_017401.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020932.mm.1 |
NM_017401.2 |
Refseq |