Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100020904.mm.1 |
TC1100002152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020904.mm.1 |
NM_018870.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020904.mm.1 |
NM_018870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020905.mm.1 |
TC1100002152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020905.mm.1 |
NM_018870.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020905.mm.1 |
NM_018870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020903.mm.1 |
TC1100002152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020903.mm.1 |
NM_018870.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020903.mm.1 |
NM_018870.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020906.mm.1 |
TC1100002152.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100020906.mm.1 |
NM_018870.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100020906.mm.1 |
NM_018870.3 |
Refseq |