Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00451399_m1
          See other RFC2 GE Assays ›
          Gene Symbol:
          Rfc2
          Gene Name:
          replication factor C (activator 1) 2
          Gene Aliases:
          2610008M13Rik, 40kDa, AI326953, Recc2
          Chromosome Location:
          Chr.5: 134582690 - 134598328 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00451399_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rfc2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020022.2 NP_064406.1 1 - 2 193 69
          GenBank mRNA AK011353.1 - 1 - 2 109 69
          AK011649.1 - 1 - 2 107 69
          AK075997.1 - 1 - 2 105 69
          AK154681.1 - 1 - 2 147 69
          BC023028.1 - 1 - 2 114 5374851 69
          BC099370.1 - 1 - 2 157 30937690 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001560.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rfc2

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          DNA-dependent DNA replication
          nucleotide binding
          DNA binding
          DNA clamp loader activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 AK154681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 BC004812.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 BC023028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 BC099370.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 NM_020022.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 AK154681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 BC004812.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 NM_020022.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 BC023028.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P254912 BC099370.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284342 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765457 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805578 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 680857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189613 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143823 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117164 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 AK154681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 BC004812.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 BC023028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 BC099370.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 NM_020022.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 AK154681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 BC004812.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 BC023028.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 BC099370.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P254912 NM_020022.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 AK154681.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 BC004812.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 BC023028.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 BC099370.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 NM_020022.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 AK154681.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 BC004812.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 BC023028.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 BC099370.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P254912 NM_020022.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117164 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09573356 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09573365 - -
          Whole Mouse Genome G4122A A_51_P254912 AK154681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254912 BC004812.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254912 BC023028.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254912 BC099370.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254912 NM_020022.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254912 AK154681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254912 BC004812.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254912 BC023028.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254912 BC099370.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P254912 NM_020022.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P981680 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106708 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P33001058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P33001057 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526218 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526225 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526220 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526228 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526227 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526226 - -
          Affymetrix HT MG-430 PM Array Plate 1457669_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1457638_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1417503_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526217 - -
          Mouse Genome 430 2.0 Array 1457669_PM_x_at - -
          Mouse Genome 430 2.0 Array 1457638_PM_x_at - -
          Mouse Genome 430 2.0 Array 1417503_PM_at - -
          Mouse Genome 430 2.0 Array 1449606_at - -
          Mouse Genome 430 2.0 Array 1457638_x_at - -
          Mouse Genome 430 2.0 Array 1449606_PM_at - -
          Mouse Genome 430 2.0 Array 1417503_at - -
          Mouse Genome 430 2.0 Array 1457669_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19718_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19718_at - -
          Affymetrix Mouse Clariom S Array PSR0500013983.mm.1 TC0500001560.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500013983.mm.1 NM_020022.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500013983.mm.1 NM_020022.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500013982.mm.1 TC0500001560.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500013982.mm.1 NM_020022.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500013982.mm.1 NM_020022.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500013981.mm.1 TC0500001560.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500013981.mm.1 NM_020022.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500013981.mm.1 NM_020022.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500013980.mm.1 TC0500001560.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500013980.mm.1 NM_020022.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500013980.mm.1 NM_020022.2 Refseq
          Mouse Expression Set 430 1417503_at - -
          Mouse Expression Set 430 1417503_PM_at - -
          Mouse Expression Set 430 1457669_x_at - -
          Mouse Expression Set 430 1457638_x_at - -
          Mouse Expression Set 430 1457638_PM_x_at - -
          Mouse Expression Set 430 1457669_PM_x_at - -
          Murine Genome U74v2 Set 160759_at - -
          Murine Genome U74v2 Set 160759_PM_at - -
          Mouse Genome 430A 2.0 Array 1417503_at - -
          Mouse Genome 430A 2.0 Array 1417503_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417503_at - -
          Affymetrix HT Mouse Genome 430A Array 1417503_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457638_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457638_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457669_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457669_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 738626 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107040121 - -
          MouseWG-6 v2 Expression BeadChip 106980288 - -
          Illumina mouse-6 v1.0 expression beadchip 106980288 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2749487 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2734977 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2424690 - -
          mouse-6 v1.1 (Illumina) scl27145.4.1_19-S - -
          mouse-6 v1.1 (Illumina) scl019718.11_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 107040121 - -
          Illumina Sentrix mouse-6.v1 bead array 106980288 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106980288 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 107040121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2894497 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2749484 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2894497 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2749487 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0