Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00451460_s1
          See other OLFR749 GE Assays ›
          Gene Symbol:
          Olfr749 Olfr747
          Gene Name:
          olfactory receptor 749
          olfactory receptor 747
          Gene Aliases:
          MOR106-1, ORL461, Ors1
          MOR106-16, MOR106-7
          Chromosome Location:
          Chr.14: 50736219 - 50744324 on Build GRCm38
          Chromosome Location:
          Chr.14: 50680691 - 50681645 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00451460_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr749
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020288.2 NP_064684.2 2 206 122
          Gene Symbol: Olfr747
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207156.2 NP_997039.2 1 140 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001983.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr749
          Click here to find siRNAs for Gene Olfr747

          Gene Ontology Categories:

          Function(s) Process(es)

          G protein-coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072591 NM_207156.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2180680 NM_020288.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827255 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133334 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072591 NM_207156.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2180680 NM_020288.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430223 NM_207156.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P430223 NM_020288.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29598207 - -
          Whole Mouse Genome G4122A A_51_P430223 NM_207156.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P430223 NM_020288.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140955 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11546834 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11546830 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11546831 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11546838 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11782950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11546833 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11782959 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11546846 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11782953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29598231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11725624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11195320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11674460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11751570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11775545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11356783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11546841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11798002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11546824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11798003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11751575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11237530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11764581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11356784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11674459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29598061 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10419459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419457 - -
          Affymetrix HT MG-430 PM Array Plate 1450589_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10419456 - -
          Affymetrix Mouse Gene 1.1 ST Array 10419458 - -
          Mouse Genome 430 2.0 Array 1450589_at - -
          Mouse Genome 430 2.0 Array 1450589_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400014839.mm.1 TC1400001983.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400014839.mm.1 NM_207156.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014839.mm.1 NM_207156.2 Refseq
          Mouse Expression Set 430 1450589_at - -
          Mouse Expression Set 430 1450589_PM_at - -
          Mouse Genome 430A 2.0 Array 1450589_at - -
          Mouse Genome 430A 2.0 Array 1450589_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450589_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450589_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 746569 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3130725 - -
          Illumina mouse-6 v1.0 expression beadchip 3130725 - -
          Illumina mouse-6 v1.0 expression beadchip 1410504 - -
          Illumina mouse-6 v1.0 expression beadchip 5670056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130725 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410504 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2721475 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224751 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232300 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2721475 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224751 - -
          mouse-6 v1.1 (Illumina) scl0258264.2_2-S - -
          mouse-6 v1.1 (Illumina) scl00258264.1_15-S - -
          mouse-6 v1.1 (Illumina) scl056858.1_193-S - -
          Illumina Sentrix mouse-6.v1 bead array 5670056 - -
          Illumina Sentrix mouse-6.v1 bead array 3130725 - -
          Illumina Sentrix mouse-6.v1 bead array 1410504 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0258264.2_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00258264.1_15-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056858.1_193-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670056 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410504 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130725 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224751 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224751 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2721475 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232300 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0