Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00451967_m1
          See other SCN5A GE Assays ›
          Gene Symbol:
          Scn5a
          Gene Name:
          sodium channel, voltage-gated, type V, alpha
          Gene Aliases:
          Nav1.5, Nav1.5c, SkM1, SkM2, mH1
          Chromosome Location:
          Chr.9: 119483404 - 119579030 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00451967_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scn5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001253860.1 NP_001240789.1 7 - 8 1052 70
          NM_021544.4 NP_067519.2 8 - 9 1220 70
          GenBank mRNA AJ271477.1 - 7 - 8 1048 70
          AK147254.1 - 8 - 9 1220 70
          AK147517.1 - 8 - 9 1220 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003244.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scn5a

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          cardiac ventricle development
          transport
          ion transport
          sodium ion transport
          positive regulation of heart rate
          positive regulation of sodium ion transport
          response to organic cyclic compound
          response to denervation involved in regulation of muscle adaptation
          neuronal action potential
          regulation of ion transmembrane transport
          sodium ion transmembrane transport
          positive regulation of action potential
          positive regulation of epithelial cell proliferation
          membrane depolarization
          transmembrane transport
          cardiac muscle contraction
          regulation of ventricular cardiac muscle cell membrane repolarization
          regulation of atrial cardiac muscle cell membrane depolarization
          regulation of atrial cardiac muscle cell membrane repolarization
          regulation of ventricular cardiac muscle cell membrane depolarization
          cellular response to calcium ion
          cardiac muscle cell action potential involved in contraction
          regulation of cardiac muscle cell contraction
          ventricular cardiac muscle cell action potential
          membrane depolarization during action potential
          membrane depolarization during cardiac muscle cell action potential
          SA node cell action potential
          AV node cell action potential
          bundle of His cell action potential
          membrane depolarization during AV node cell action potential
          membrane depolarization during SA node cell action potential
          membrane depolarization during Purkinje myocyte cell action potential
          membrane depolarization during bundle of His cell action potential
          AV node cell to bundle of His cell communication
          regulation of heart rate by cardiac conduction
          regulation of sodium ion transmembrane transport
          ion channel activity
          voltage-gated ion channel activity
          voltage-gated sodium channel activity
          sodium channel activity
          protein binding
          calmodulin binding
          fibroblast growth factor binding
          enzyme binding
          protein kinase binding
          ankyrin binding
          ubiquitin protein ligase binding
          transmembrane transporter binding
          nitric-oxide synthase binding
          voltage-gated sodium channel activity involved in cardiac muscle cell action potential
          voltage-gated sodium channel activity involved in AV node cell action potential
          voltage-gated sodium channel activity involved in bundle of His cell action potential
          voltage-gated sodium channel activity involved in Purkinje myocyte action potential
          voltage-gated sodium channel activity involved in SA node cell action potential
          scaffold protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900027317.mm.1 TC0900003244.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900027317.mm.1 NM_001253860.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027317.mm.1 NM_001253860.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027317.mm.1 NM_021544.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027317.mm.1 NM_021544.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0