Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00453011_m1
          See other SCPEP1 GE Assays ›
          Gene Symbol:
          Scpep1
          Gene Name:
          serine carboxypeptidase 1
          Gene Aliases:
          2410018F01Rik, 4833411K15Rik, AI957256, Risc
          Chromosome Location:
          Chr.11: 88924020 - 88955442 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00453011_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scpep1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029023.3 NP_083299.3 11 - 12 1198 70
          GenBank mRNA AF330052.1 - 11 - 12 1154 70
          AK014680.1 - 11 - 12 1199 70
          AK050181.1 - 11 - 12 1136 70
          AK153225.1 - 11 - 12 1186 70
          AK153485.1 - 10 - 11 987 70
          AK190943.1 - 1 - 2 73 70
          BC004847.1 - 11 - 12 1188 2650587 70
          BC021399.1 - 11 - 12 1153 4193674 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003506.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scpep1

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          negative regulation of blood pressure
          carboxypeptidase activity
          serine-type carboxypeptidase activity
          peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28003084 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28003085 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28003082 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990210 BC021399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990210 BC004847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P136714 BC021399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P136714 BC004847.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566330 AK033940.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791078 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 650950 NM_001011737.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 137324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249574 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 882375 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 550681 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861826 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79641 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174119 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121589 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990210 BC021399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990210 BC004847.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118417 BC021399.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118417 BC004847.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118415 BC021399.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P118415 BC004847.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28003086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28003091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28003103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28003084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28003093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28003089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28003078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28003076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28003081 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121589 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02389710 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02389702 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02389743 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07435026 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174119 - -
          Whole Mouse Genome G4122A A_51_P118417 BC021399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118417 BC004847.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118415 BC021399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P118415 BC004847.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136714 BC021399.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136714 BC004847.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02389743 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174119 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09393055 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09393056 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09393060 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28003085 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28003082 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28003084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28002986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28003046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09069690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09490293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09063995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09526716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09547370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09490294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09335394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09335392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09393055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09445389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28003081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09421262 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10389724 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389731 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389720 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389723 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389730 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389728 - -
          Affymetrix Mouse Gene 1.0 ST Array 10389729 - -
          Affymetrix HT MG-430 PM Array Plate 1426555_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455908_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10389719 - -
          Mouse Genome 430 2.0 Array 1426555_PM_at - -
          Mouse Genome 430 2.0 Array 1455908_PM_a_at - -
          Mouse Genome 430 2.0 Array 1426555_at - -
          Mouse Genome 430 2.0 Array 1455908_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74617_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74617_at - -
          Affymetrix Mouse Clariom S Array PSR1100032092.mm.1 TC1100003506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032092.mm.1 NM_029023.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032092.mm.1 NM_029023.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032093.mm.1 TC1100003506.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100032093.mm.1 NM_029023.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100032093.mm.1 NM_029023.3 Refseq
          Mouse Expression Set 430 1455908_PM_a_at - -
          Mouse Expression Set 430 1455908_a_at - -
          Mouse Expression Set 430 1426555_at - -
          Mouse Expression Set 430 1426555_PM_at - -
          Murine Genome U74v2 Set 98042_PM_at - -
          Murine Genome U74v2 Set 98042_at - -
          Mouse Genome 430A 2.0 Array 1455908_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426555_PM_at - -
          Mouse Genome 430A 2.0 Array 1426555_at - -
          Mouse Genome 430A 2.0 Array 1455908_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426555_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455908_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1455908_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426555_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3610576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2736968 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2736968 - -
          mouse-6 v1.1 (Illumina) scl39738.13.1_56-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl39738.13.1_56-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2736968 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2736968 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0