Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00455118_m1
          See other SLC2A9 GE Assays ›
          Gene Symbol:
          Slc2a9
          Gene Name:
          solute carrier family 2 (facilitated glucose transporter), member 9
          Gene Aliases:
          Glut9, SLC2A9B, SLC2a9A
          Chromosome Location:
          Chr.5: 38349273 - 38503149 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00455118_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc2a9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012363.2 NP_001012363.2 2 - 3 169 81
          NM_001102414.1 NP_001095884.1 2 - 3 368 81
          NM_001102415.1 NP_001095885.1 2 - 3 368 81
          NM_145559.2 NP_663534.1 3 - 4 363 81
          GenBank mRNA AF469480.1 - 2 - 3 372 81
          AF490463.1 - 2 - 3 193 81
          AK037093.1 - 2 - 3 221 81
          AK156740.1 - 2 - 3 272 81
          AK172234.1 - 2 - 3 272 81
          AY776155.1 - 2 - 3 148 81
          BC006076.1 - 3 - 4 363 3593581 81
          BC138213.1 - 3 - 4 304 8861232 81
          BC145198.1 - 3 - 4 304 9053749 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002284.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc2a9

          Gene Ontology Categories:

          Function(s) Process(es)

          hexose transmembrane transport
          carbohydrate transmembrane transport
          glucose import
          urate metabolic process
          cation transmembrane transport
          proton transmembrane transport
          glucose transmembrane transport
          carbohydrate:proton symporter activity
          glucose transmembrane transporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500021081.mm.1 TC0500002284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500021081.mm.1 NM_001012363.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021081.mm.1 NM_001012363.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021081.mm.1 NM_001102414.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021081.mm.1 NM_001102414.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021081.mm.1 NM_001102415.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021081.mm.1 NM_001102415.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021081.mm.1 NM_145559.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500021081.mm.1 NM_145559.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c8579b687-54cxl:80/100.66.135.11:80.
          git-commit: 8e87205ac1279514f9c7a0d34bcd4a43061862b7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0