Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00456071_m1
          See other BSCL2 GE Assays ›
          Gene Symbol:
          Bscl2
          Gene Name:
          Berardinelli-Seip congenital lipodystrophy 2 homolog (seipin)
          Gene Aliases:
          2900097C17Rik, AI046355, Gng3lg
          Chromosome Location:
          Chr.19: 8837467 - 8848683 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00456071_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bscl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001136064.3 NP_001129536.1 2 - 3 606 71
          NM_008144.5 NP_032170.3 1 - 2 456 71
          GenBank mRNA AB030196.1 - 1 - 2 75 71
          AF069954.2 - 1 - 2 281 71
          AK158901.1 - 1 - 2 457 71
          BC043023.1 - 1 - 2 220 5689013 71
          BC060048.1 - 1 - 2 411 71
          BC061689.1 - 2 - 3 344 6306706 71

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AB030196.1, AF069954.2, BC043023.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bscl2

          Gene Ontology Categories:

          Function(s)

          lipid metabolic process
          lipid catabolic process
          lipid storage
          lipid droplet organization
          fat cell differentiation
          negative regulation of lipid catabolic process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 BC043023.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 BC060048.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 BC061689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 AK158901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 AK170183.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 BC043023.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 BC060048.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 BC061689.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 AK158901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P196526 AK170183.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968447 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 797863 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 158978 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111792 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173734 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 BC043023.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 BC060048.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 BC061689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 AK158901.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 AK170183.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 BC043023.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 BC060048.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 BC061689.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 AK158901.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P196526 AK170183.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P256584 CK618551.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P256584 BC043023.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P256584 BC060048.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P256584 BC061689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P256584 AK158901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 BC043023.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 BC060048.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 BC061689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 AK158901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 AK170183.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 BC043023.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 BC060048.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 BC061689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 AK158901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196526 AK170183.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121686 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14232021 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14232020 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14232018 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461250 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461249 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461244 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461246 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461242 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461241 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461248 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461247 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461240 - -
          Affymetrix HT MG-430 PM Array Plate 1420632_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1437345_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461237 - -
          Mouse Genome 430 2.0 Array 1437345_a_at - -
          Mouse Genome 430 2.0 Array 1420632_a_at - -
          Mouse Genome 430 2.0 Array 1420632_PM_a_at - -
          Mouse Genome 430 2.0 Array 1437345_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14705_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14705_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900002121.mm.1 TC1900000160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002121.mm.1 NM_001136064.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002122.mm.1 TC1900000160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002122.mm.1 NM_001136064.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002119.mm.1 TC1900000160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002119.mm.1 NM_001136064.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002127.mm.1 TC1900000160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002127.mm.1 NM_001136064.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002127.mm.1 NM_008144.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002133.mm.1 TC1900000160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002133.mm.1 NM_001136064.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002133.mm.1 NM_008144.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002125.mm.1 TC1900000160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002125.mm.1 NM_008144.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900002123.mm.1 TC1900000160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900002123.mm.1 NM_008144.5 Refseq
          Mouse Expression Set 430 1420632_a_at - -
          Mouse Expression Set 430 1437345_PM_a_at - -
          Mouse Expression Set 430 1437345_a_at - -
          Mouse Expression Set 430 1420632_PM_a_at - -
          Murine Genome U74v2 Set 93080_PM_at - -
          Murine Genome U74v2 Set 93080_at - -
          Mouse Genome 430A 2.0 Array 1437345_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1420632_a_at - -
          Mouse Genome 430A 2.0 Array 1437345_a_at - -
          Mouse Genome 430A 2.0 Array 1420632_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1420632_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437345_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437345_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1420632_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 308943 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2680671 - -
          Illumina mouse-6 v1.0 expression beadchip 2680671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680671 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221719 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221719 - -
          mouse-6 v1.1 (Illumina) scl53403.26.13_76-S - -
          Illumina Sentrix mouse-6.v1 bead array 2680671 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl53403.26.13_76-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680671 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1221719 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221719 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl53403.26.13_76-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0