Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00457046_m1
          See other TBPL1 GE Assays ›
          Gene Symbol:
          Tbpl1
          Gene Name:
          TATA box binding protein-like 1
          Gene Aliases:
          4732475G08, AW011832, AW491032, D18347, STD, TLF, TRF2, TRP, Tlp
          Chromosome Location:
          Chr.10: 22703877 - 22732573 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00457046_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbpl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011603.5 NP_035733.1 1 - 2 331 99
          GenBank mRNA AB017697.1 - 1 - 2 228 99
          AK049975.1 - 1 - 2 203 99
          AK078547.1 - 1 - 2 263 99
          BC033926.1 - 1 - 2 172 1382180 99

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AB017697.1, AK049975.1, AK078547.1, BC033926.1, NM_011603.5 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tbpl1

          Gene Ontology Categories:

          Function(s) Process(es)

          acrosome assembly
          dTTP biosynthetic process
          transcription, DNA-templated
          DNA-templated transcription, initiation
          regulation of transcription, DNA-templated
          spermatogenesis
          spermatid nucleus differentiation
          DNA binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2018442 BC033926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 BC033926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 AK005604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 AK049975.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 AK078547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 NM_011603.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 BC033926.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 AK005604.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 AK049975.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 AK078547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P422629 NM_011603.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966346 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322226 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57853 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153652 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102279 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2018442 BC033926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 BC033926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 AK005604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 AK049975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 AK078547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 NM_011603.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 BC033926.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 AK005604.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 AK049975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 AK078547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P422629 NM_011603.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 BC033926.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 AK005604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 AK049975.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 AK078547.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 NM_011603.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 BC033926.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 AK005604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 AK049975.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 AK078547.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422629 NM_011603.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P64924 BE650743.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P64924 AK049975.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P64924 AK078547.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P64924 NM_011603.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422636 BC033926.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422636 AK005604.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422636 AK049975.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422636 AK078547.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P422636 NM_011603.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 BC033926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 AK005604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 AK049975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 AK078547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 NM_011603.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 BC033926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 AK005604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 AK049975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 AK078547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422629 NM_011603.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P387690 - -
          Whole Mouse Genome G4122A A_52_P64924 BE650743.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64924 AK049975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64924 AK078547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64924 NM_011603.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422636 BC033926.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422636 AK005604.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422636 AK049975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422636 AK078547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P422636 NM_011603.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127820 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115052 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08645263 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08645264 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08950407 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08645261 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08645277 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08950405 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10368231 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368236 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368235 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368232 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368237 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368234 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368233 - -
          Affymetrix HT MG-430 PM Array Plate 1431415_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1434907_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10368229 - -
          Mouse Genome 430 2.0 Array 1434907_at - -
          Mouse Genome 430 2.0 Array 1434907_PM_at - -
          Mouse Genome 430 2.0 Array 1431415_a_at - -
          Mouse Genome 430 2.0 Array 1431415_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237336_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237336_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000013458.mm.1 TC1000001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013458.mm.1 NM_011603.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013458.mm.1 NM_011603.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013459.mm.1 TC1000001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013459.mm.1 NM_011603.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013459.mm.1 NM_011603.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013456.mm.1 TC1000001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013456.mm.1 NM_011603.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013456.mm.1 NM_011603.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013457.mm.1 TC1000001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013457.mm.1 NM_011603.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013457.mm.1 NM_011603.5 Refseq
          Mouse Expression Set 430 1431415_PM_a_at - -
          Mouse Expression Set 430 1431415_a_at - -
          Mouse Expression Set 430 1434907_at - -
          Mouse Expression Set 430 1434907_PM_at - -
          Murine Genome U74v2 Set 97893_at - -
          Murine Genome U74v2 Set 97893_PM_at - -
          Mouse Genome 430A 2.0 Array 1431415_a_at - -
          Mouse Genome 430A 2.0 Array 1431415_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1431415_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1431415_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1434907_at - -
          Affymetrix HT Mouse Genome 430B Array 1434907_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 353530 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3610332 - -
          Illumina mouse-6 v1.0 expression beadchip 3610332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605872 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605872 - -
          mouse-6 v1.1 (Illumina) scl38128.9_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38128.9_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605872 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605872 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline