Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200008612.mm.1 |
TC0200001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008612.mm.1 |
NM_001164791.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008612.mm.1 |
NM_001164791.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008612.mm.1 |
NM_023057.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008612.mm.1 |
NM_023057.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008612.mm.1 |
NM_178084.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008612.mm.1 |
NM_178084.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008613.mm.1 |
TC0200001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008613.mm.1 |
NM_001164791.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008613.mm.1 |
NM_001164791.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008613.mm.1 |
NM_023057.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008613.mm.1 |
NM_023057.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008613.mm.1 |
NM_178084.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008613.mm.1 |
NM_178084.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008614.mm.1 |
TC0200001053.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200008614.mm.1 |
NM_001164791.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008614.mm.1 |
NM_001164791.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008614.mm.1 |
NM_023057.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008614.mm.1 |
NM_023057.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008614.mm.1 |
NM_178084.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200008614.mm.1 |
NM_178084.4 |
Refseq |