Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023603.3 | NP_076092.1 | 2 - 3 | 1111 | 63 | |
NR_045010.1 | 2 - 3 | 1111 | 63 | |||
GenBank mRNA | AF272847.1 | - | 2 - 3 | 404 | 63 | |
AK157783.1 | - | 3 - 4 | 748 | 63 | ||
AK211570.1 | - | 1 - 2 | 51 | 63 | ||
AY034062.1 | - | 2 - 3 | 1026 | 63 | ||
BC080848.1 | - | 2 - 3 | 1077 | 63 | ||
BC089305.1 | - | 2 - 3 | 1077 | 30360144 | 63 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |