Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AU040320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035526.1 | NP_001030603.1 | 1 - 2 | 223 | 54 | |
NM_133886.2 | NP_598647.1 | 1 - 2 | 223 | 54 | ||
GenBank mRNA | AK031342.1 | - | 1 - 2 | 167 | 54 | |
AK043006.1 | - | 1 - 2 | 173 | 54 | ||
AK170261.1 | - | 1 - 2 | 180 | 54 | ||
BC028869.1 | - | 1 - 2 | 193 | 4235813 | 54 | |
BY289654.1 | - | 1 - 2 | 224 | 54 | ||
BY308757.1 | - | 1 - 2 | 199 | 54 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |