Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AU040320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035525.1 | NP_001030602.1 | 17 - 18 | 2788 | 64 | |
NM_001035526.1 | NP_001030603.1 | 17 - 18 | 2906 | 64 | ||
NM_133886.2 | NP_598647.1 | 17 - 18 | 2906 | 64 | ||
GenBank mRNA | AK031342.1 | - | 17 - 18 | 2850 | 64 | |
AK043006.1 | - | 17 - 18 | 2856 | 64 | ||
AK084668.1 | - | 17 - 18 | 2751 | 64 | ||
AK147569.1 | - | 5437 | 64 | |||
AK161493.1 | - | 7 - 8 | 969 | 64 | ||
AK170261.1 | - | 17 - 18 | 2863 | 64 | ||
BC022154.1 | - | 8 - 9 | 1318 | 64 | ||
BC028869.1 | - | 17 - 18 | 2876 | 4235813 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |