Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162868.1 | NP_001156340.1 | 4 - 5 | 1998 | 74 | |
NM_001162869.1 | NP_001156341.1 | 4 - 5 | 1998 | 74 | ||
NM_153140.2 | NP_694780.1 | 2 - 3 | 389 | 74 | ||
GenBank mRNA | AB093257.1 | - | 4 - 5 | 1439 | 74 | |
AK052775.1 | - | 2 - 3 | 348 | 74 | ||
AK083658.1 | - | 4 - 5 | 1682 | 74 | ||
AK085516.1 | - | 2 - 3 | 390 | 74 | ||
AK128985.1 | - | 2 - 3 | 389 | 74 | ||
BC023364.1 | - | 2 - 3 | 380 | 74 | ||
BC037132.1 | - | 2 - 3 | 429 | 74 | ||
CF170100.1 | - | 2 - 3 | 288 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |