Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165983.1 | NP_001159455.1 | 15 - 16 | 1866 | 103 | |
NM_001165984.1 | NP_001159456.1 | 15 - 16 | 1826 | 103 | ||
NM_001165985.1 | NP_001159457.1 | 15 - 16 | 1884 | 103 | ||
NM_001165986.1 | NP_001159458.1 | 15 - 16 | 1859 | 103 | ||
NM_001165987.1 | NP_001159459.1 | 14 - 15 | 1791 | 103 | ||
NM_001165988.1 | NP_001159460.1 | 14 - 15 | 1766 | 103 | ||
NM_028475.2 | NP_082751.1 | 15 - 16 | 1851 | 103 | ||
NM_153489.2 | NP_705693.2 | 14 - 15 | 1791 | 103 | ||
GenBank mRNA | AF276967.1 | - | 14 - 15 | 1726 | 103 | |
AK016538.1 | - | 8 - 9 | 1118 | 103 | ||
AK030218.1 | - | 15 - 16 | 1792 | 103 | ||
AK031934.1 | - | 14 - 15 | 1771 | 103 | ||
AK048286.1 | - | 15 - 16 | 1867 | 103 | ||
AK079895.1 | - | 15 - 16 | 1823 | 103 | ||
AK088702.1 | - | 15 - 16 | 1882 | 103 | ||
AK122210.1 | - | 2 - 3 | 229 | 103 | ||
BC023906.1 | - | 14 - 15 | 1782 | 5321969 | 103 | |
BC029075.1 | - | 15 - 16 | 1841 | 4527901 | 103 | |
BC050910.1 | - | 14 - 15 | 1773 | 6417527 | 103 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |