Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00469062_m1
          See other ENO2 GE Assays ›
          Gene Symbol:
          Eno2
          Gene Name:
          enolase 2, gamma neuronal
          Gene Aliases:
          AI837106, D6Ertd375e, Eno-2, NSE
          Chromosome Location:
          Chr.6: 124760053 - 124769673 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00469062_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eno2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK003051.1 - 7 - 8 761 76
          AK131652.1 - 7 - 8 800 76
          AK134571.1 - 6 - 7 684 76
          AK146655.1 - 7 - 8 766 76
          AK170169.1 - 6 - 7 681 76
          BC031739.1 - 7 - 8 754 4527274 76
          X52380.1 - 7 - 8 779 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600003116.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816933 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816931 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816932 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019457 BC043708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019457 X52380.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019457 BC009018.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019457 BC031739.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2019457 AK146655.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 747616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 145215 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170901 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019457 BC043708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019457 X52380.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019457 BC009018.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019457 BC031739.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2019457 AK146655.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P130028 BC043708.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P130028 X52380.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P130028 BC009018.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P130028 BC031739.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P130028 AK146655.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P748882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24816915 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06056303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06056292 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170901 - -
          Whole Mouse Genome G4122A A_52_P748882 - -
          Whole Mouse Genome G4122A A_51_P130028 BC043708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P130028 X52380.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P130028 BC009018.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P130028 BC031739.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P130028 AK146655.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114691 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05688755 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816933 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816931 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24816932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24816915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05597491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05817355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05817352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05597481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05817360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05410237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05410241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24816898 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547813 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547812 - -
          Affymetrix HT MG-430 PM Array Plate 1418829_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10547807 - -
          Mouse Genome 430 2.0 Array 1418829_PM_a_at - -
          Mouse Genome 430 2.0 Array 1418829_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13807_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13807_at - -
          Affymetrix Mouse Clariom S Array PSR0600024166.mm.1 TC0600003116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024166.mm.1 NM_013509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024171.mm.1 TC0600003116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024171.mm.1 NM_013509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024170.mm.1 TC0600003116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024170.mm.1 NM_013509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024167.mm.1 TC0600003116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024167.mm.1 NM_013509.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600024169.mm.1 TC0600003116.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600024169.mm.1 NM_013509.3 Refseq
          Mouse Expression Set 430 1418829_PM_a_at - -
          Mouse Expression Set 430 1418829_a_at - -
          Murine Genome U74v2 Set 99045_at - -
          Murine Genome U74v2 Set 99045_PM_at - -
          Mouse Genome 430A 2.0 Array 1418829_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1418829_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1418829_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1418829_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 482566 - -
          ABI Mouse Genome Survey Microarray 926113 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320068 - -
          Illumina mouse-6 v1.0 expression beadchip 2320068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320068 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2763002 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2763002 - -
          mouse-6 v1.1 (Illumina) scl013807.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 2320068 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013807.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2763002 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2763002 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl013807.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6cbc4fbdf-xtbd4:80/100.66.128.131:80.
          git-commit: 26a732473c29a7560faabcf99da408bd94702681
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc