Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00474585_s1
          See other PCDHB19 GE Assays ›
          Gene Symbol:
          Pcdhb19
          Gene Name:
          protocadherin beta 19
          Gene Aliases:
          Pcdhb11, PcdhbS
          Chromosome Location:
          Chr.18: 37496998 - 37501711 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00474585_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdhb19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053144.2 NP_444374.2 1 2767 95
          GenBank mRNA AF326312.1 - 1 2611 95
          AK029163.1 - 1 2775 95
          AK046363.1 - 1 2768 95
          AK147520.1 - 2770 95
          AK160654.1 - 1 2769 96
          BC141290.1 - 1 2686 9055902 95
          BC141291.1 - 1 2686 9055903 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000376.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcdhb19

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P252953 AK046363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P252953 AK147520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P252953 NM_053144.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P252953 AK046363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P252953 AK147520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P252953 NM_053144.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 695786 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184730 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155471 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P252953 AK046363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P252953 AK147520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P252953 NM_053144.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P252953 AK046363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P252953 AK147520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P252953 NM_053144.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268469 BC141291.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268469 BC141290.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268469 AK029163.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268469 AK046363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P268469 AK147520.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P252953 AK046363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P252953 AK147520.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P252953 NM_053144.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P252953 AK046363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P252953 AK147520.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P252953 NM_053144.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628606 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155471 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08968331 - -
          Whole Mouse Genome G4122A A_52_P252953 AK046363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252953 AK147520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252953 NM_053144.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252953 AK046363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252953 AK147520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252953 NM_053144.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268469 BC141291.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268469 BC141290.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268469 AK029163.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268469 AK046363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P268469 AK147520.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139197 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155471 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13915312 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13915356 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13915319 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13915338 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13915327 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13915318 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13915310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13915333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13915320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14025026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628612 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455126 - -
          Affymetrix HT MG-430 PM Array Plate 1420443_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455123 - -
          Mouse Genome 430 2.0 Array 1420443_at - -
          Mouse Genome 430 2.0 Array 1420443_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93890_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93890_at - -
          Affymetrix Mouse Clariom S Array PSR1800002869.mm.1 TC1800000376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002869.mm.1 NM_053144.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002869.mm.1 NM_053144.2 Refseq
          Mouse Expression Set 430 1420443_PM_at - -
          Mouse Expression Set 430 1420443_at - -
          Mouse Genome 430A 2.0 Array 1420443_at - -
          Mouse Genome 430A 2.0 Array 1420443_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420443_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420443_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105900576 - -
          Illumina mouse-6 v1.0 expression beadchip 2190411 - -
          Illumina mouse-6 v1.0 expression beadchip 105900576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2747391 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257036 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2747391 - -
          mouse-6 v1.1 (Illumina) scl52054.1_546-S - -
          mouse-6 v1.1 (Illumina) scl0093890.2_329-S - -
          Illumina Sentrix mouse-6.v1 bead array 105900576 - -
          Illumina Sentrix mouse-6.v1 bead array 2190411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0093890.2_329-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105900576 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190411 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2465874 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2465874 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0