Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195031.1 | NP_001181960.1 | 5 - 6 | 861 | 74 | |
NM_053182.5 | NP_444412.2 | 4 - 5 | 802 | 74 | ||
GenBank mRNA | AF250192.1 | - | 2 - 3 | 74 | ||
AK040230.1 | - | 5 - 6 | 811 | 74 | ||
AK076515.1 | - | 1 - 2 | 300 | 74 | ||
AK089329.1 | - | 4 - 5 | 754 | 74 | ||
AK156000.1 | - | 5 - 6 | 813 | 74 | ||
AK156986.1 | - | 5 - 6 | 786 | 74 | ||
AK172106.1 | - | 5 - 6 | 784 | 74 | ||
BC145328.1 | - | 5 - 6 | 685 | 9053884 | 74 | |
BC145761.1 | - | 5 - 6 | 685 | 40131064 | 74 | |
BC145763.1 | - | 5 - 6 | 685 | 40131066 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |