Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00476806_m1
          See other PIP5K1C GE Assays ›
          Gene Symbol:
          Pip5k1c
          Gene Name:
          phosphatidylinositol-4-phosphate 5-kinase, type 1 gamma
          Gene Aliases:
          AI115456, AI835305, Pip5kIgamma
          Chromosome Location:
          Chr.10: 81292962 - 81319974 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00476806_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pip5k1c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146687.2 NP_001140159.1 5 - 6 565 70
          NM_001293646.1 NP_001280575.1 5 - 6 565 70
          NM_001293647.1 NP_001280576.1 5 - 6 578 70
          NM_008844.3 NP_032870.2 5 - 6 565 70
          GenBank mRNA AB006916.1 - 5 - 6 548 70
          AK014403.1 - 5 - 6 531 70
          AK041402.1 - 5 - 6 551 70
          AK122319.2 - 5 - 6 523 70
          AK154816.1 - 5 - 6 557 70
          AK155371.1 - 5 - 6 540 70
          AK170286.1 - 5 - 6 557 70
          AK171576.1 - 5 - 6 539 70
          BC019138.1 - 5 - 6 514 5041431 70
          BC020365.1 - 5 - 6 527 70
          BC094665.1 - 5 - 6 537 30356181 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000892.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pip5k1c

          Gene Ontology Categories:

          Function(s) Process(es)

          exocytosis
          endocytosis
          phagocytosis
          chemotaxis
          cell adhesion
          axonogenesis
          phosphorylation
          phosphatidylinositol metabolic process
          phosphatidylinositol phosphate biosynthetic process
          platelet aggregation
          nucleotide binding
          protein binding
          ATP binding
          kinase activity
          phosphatidylinositol phosphate kinase activity
          1-phosphatidylinositol-4-phosphate 5-kinase activity
          transferase activity
          talin binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000007003.mm.1 TC1000000892.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007003.mm.1 NM_001146687.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007003.mm.1 NM_008844.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007005.mm.1 TC1000000892.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007005.mm.1 NM_001146687.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007005.mm.1 NM_008844.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007004.mm.1 TC1000000892.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000007004.mm.1 NM_001146687.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000007004.mm.1 NM_008844.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0