Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00480246_m1
          See other ALG2 GE Assays ›
          Gene Symbol:
          Alg2
          Gene Name:
          asparagine-linked glycosylation 2 (alpha-1,3-mannosyltransferase)
          Gene Aliases:
          1110018A23Rik, 1300013N08Rik, ALPG2, CDGIi, MNCb-5081
          Chromosome Location:
          Chr.4: 47469833 - 47474374 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00480246_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AB041604.1 - 2 - 3 1308 93

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22894884 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22894883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22894882 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965674 BC052411.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965674 BG093758.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965674 BC051951.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965674 AK010673.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965674 AK003764.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566735 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107078 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965674 BC052411.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965674 BG093758.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965674 BC051951.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965674 AK010673.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965674 AK003764.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123060 BC052411.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123060 BG093758.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123060 BC051951.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123060 AK010673.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P123060 AB041604.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22894877 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107078 - -
          Whole Mouse Genome G4122A A_51_P123060 BC052411.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123060 BG093758.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123060 BC051951.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123060 AK010673.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P123060 AB041604.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113452 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107078 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03605519 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03605510 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03239591 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03605507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03605502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22894883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22894884 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22894882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22894825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03537113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03379721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03395075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04083670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03938882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04083659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03605510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03537124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04083669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03395070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03395077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03605519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04083653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22894854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03537115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04083671 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512848 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512849 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512850 - -
          Affymetrix HT MG-430 PM Array Plate 1433144_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1421059_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512847 - -
          Mouse Genome 430 2.0 Array 1433144_at - -
          Mouse Genome 430 2.0 Array 1421059_a_at - -
          Mouse Genome 430 2.0 Array 1421059_PM_a_at - -
          Mouse Genome 430 2.0 Array 1433144_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56737_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56737_PM_at - -
          Mouse Expression Set 430 1421059_a_at - -
          Mouse Expression Set 430 1421059_PM_a_at - -
          Murine Genome U74v2 Set 104277_PM_at - -
          Murine Genome U74v2 Set 104277_at - -
          Mouse Genome 430A 2.0 Array 1421059_a_at - -
          Mouse Genome 430A 2.0 Array 1421059_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1433144_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421059_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421059_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1433144_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3990091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003990091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2770492 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2770492 - -
          mouse-6 v1.1 (Illumina) scl24346.2_118-S - -
          Illumina Sentrix mouse-6.v1 bead array 3990091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24346.2_118-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3990091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2803334 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2770492 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2803334 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl24346.2_118-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0