Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00480952_m1
          See other CHTOP GE Assays ›
          Gene Symbol:
          Chtop
          Gene Name:
          chromatin target of PRMT1
          Gene Aliases:
          2500003M10Rik, Fop, Srag
          Chromosome Location:
          Chr.3: 90498539 - 90509888 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00480952_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chtop
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001293776.1 NP_001280705.1 1 - 2 308 118
          NM_001293777.1 NP_001280706.1 1 - 2 308 118
          NM_001293778.1 NP_001280707.1 1 - 2 308 118
          NM_001293779.1 NP_001280708.1 1 - 2 308 118
          NM_001293780.1 NP_001280709.1 1 - 2 308 118
          NM_001293781.1 NP_001280710.1 1 - 2 308 118
          NM_001293782.1 NP_001280711.1 1 - 2 308 118
          NM_023215.6 NP_075704.2 1 - 2 308 118
          GenBank mRNA AB041654.1 - 1 - 2 260 118
          AK004558.2 - 1 - 2 254 118
          AK005377.1 - 1 - 2 273 118
          AK010866.1 - 1 - 2 290 118
          AK047914.1 - 1 - 2 282 118
          AK077960.1 - 1 - 2 297 118
          AK132769.1 - 1 - 2 281 118
          AK135914.1 - 1 - 2 285 118
          AK152492.1 - 1 - 2 261 118
          BC051939.1 - 1 - 2 262 118
          BC066837.1 - 1 - 2 261 118
          BQ746440.1 - 1 - 2 262 118
          CX234428.1 - 1 - 2 151 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002373.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AB041654.1, AK004558.2, AK005377.1, AK010866.1, AK047914.1, AK077960.1, AK132769.1, AK152492.1, NM_001293776.1, NM_001293777.1, NM_001293778.1, NM_001293779.1, NM_001293780.1, NM_001293781.1, NM_001293782.1, NM_023215.6 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chtop

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          RNA binding
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300018493.mm.1 TC0300002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018493.mm.1 NM_023215.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018495.mm.1 TC0300002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018495.mm.1 NM_023215.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018498.mm.1 TC0300002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018498.mm.1 NM_023215.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018494.mm.1 TC0300002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018494.mm.1 NM_023215.6 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018496.mm.1 TC0300002373.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018496.mm.1 NM_023215.6 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 600552 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-746d97c75b-2cnrv:80/100.66.135.11:80.
          git-commit: a7e96c5202fecf029dfaed736aa540acb41e6c9b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/PDP_Cmrc_Card_Notification_Changes