Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481091_s1
          See other CHMP1B GE Assays ›
          Gene Symbol:
          Chmp1b
          Gene Name:
          charged multivesicular body protein 1B
          Gene Aliases:
          2810405I11Rik, Chmp1b1
          Chromosome Location:
          Chr.18: 67205359 - 67207887 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481091_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chmp1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_024190.2 NP_077152.1 1 115 102
          GenBank mRNA AK012996.1 - 1 65 102
          AK014091.1 - 1 82 102
          AK151071.1 - 1 84 102
          AK167614.1 - 1 54 102
          BC002229.1 - 1 51 3491354 102

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK014091.1, AK151071.1, BC002229.1, NM_024190.2 transcripts.
          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chmp1b

          Gene Ontology Categories:

          Function(s) Process(es)

          septum digestion after cytokinesis
          transport
          nucleus organization
          vacuolar transport
          cell cycle
          mitotic metaphase plate congression
          regulation of centrosome duplication
          protein transport
          viral budding via host ESCRT complex
          establishment of protein localization
          cell division
          regulation of mitotic spindle assembly
          protein domain specific binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790570 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790564 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790566 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790565 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790572 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790567 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790569 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 BC034442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 BC002229.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 AK014091.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 AK151071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 AK167614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 BC034442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 BC002229.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 AK014091.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 AK151071.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P338110 AK167614.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P176960 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 BC034442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 BC002229.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 AK014091.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 AK151071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 AK167614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 BC034442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 BC002229.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 AK014091.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 AK151071.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P338110 AK167614.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31790543 - -
          Whole Mouse Genome G4122A A_52_P338110 BC034442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338110 BC002229.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338110 AK014091.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338110 AK151071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338110 AK167614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338110 BC034442.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338110 BC002229.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338110 AK014091.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338110 AK151071.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P338110 AK167614.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14070659 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14070667 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14070661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14070668 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14070664 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14070673 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790564 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790572 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790569 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790570 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790566 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790567 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31790565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31790566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13819381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13764864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14135198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13944963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14070667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14070673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14135195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14057091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31790575 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10456381 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456380 - -
          Affymetrix HT MG-430 PM Array Plate 1418817_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418816_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10456378 - -
          Mouse Genome 430 2.0 Array 1418817_PM_at - -
          Mouse Genome 430 2.0 Array 1446491_at - -
          Mouse Genome 430 2.0 Array 1446491_PM_at - -
          Mouse Genome 430 2.0 Array 1418817_at - -
          Mouse Genome 430 2.0 Array 1418816_PM_at - -
          Mouse Genome 430 2.0 Array 1418816_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67064_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67064_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800005134.mm.1 TC1800000698.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005134.mm.1 NM_024190.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005134.mm.1 NM_024190.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005135.mm.1 TC1800000698.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005135.mm.1 NM_024190.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005135.mm.1 NM_024190.2 Refseq
          Mouse Expression Set 430 1418816_PM_at - -
          Mouse Expression Set 430 1418817_at - -
          Mouse Expression Set 430 1418817_PM_at - -
          Mouse Expression Set 430 1418816_at - -
          Murine Genome U74v2 Set 161513_PM_r_at - -
          Murine Genome U74v2 Set 161513_r_at - -
          Mouse Genome 430A 2.0 Array 1418816_PM_at - -
          Mouse Genome 430A 2.0 Array 1418817_PM_at - -
          Mouse Genome 430A 2.0 Array 1418816_at - -
          Mouse Genome 430A 2.0 Array 1418817_at - -
          Affymetrix HT Mouse Genome 430A Array 1418816_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418817_at - -
          Affymetrix HT Mouse Genome 430A Array 1418816_at - -
          Affymetrix HT Mouse Genome 430A Array 1418817_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 680222 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102450341 - -
          MouseWG-6 v2 Expression BeadChip 3290239 - -
          MouseWG-6 v2 Expression BeadChip 106980408 - -
          Illumina mouse-6 v1.0 expression beadchip 106980408 - -
          Illumina mouse-6 v1.0 expression beadchip 102450341 - -
          Illumina mouse-6 v1.0 expression beadchip 3290239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222607 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245454 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213823 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222607 - -
          mouse-6 v1.1 (Illumina) scl51835.3_316-S - -
          mouse-6 v1.1 (Illumina) scl14871.1.1_52-S - -
          mouse-6 v1.1 (Illumina) scl0067064.2_260-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290239 - -
          Illumina Sentrix mouse-6.v1 bead array 102450341 - -
          Illumina Sentrix mouse-6.v1 bead array 106980408 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067064.2_260-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290239 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102450341 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106980408 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222607 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213823 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222607 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0067064.2_260-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0