Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481570_g1
          See other COPRS GE Assays ›
          Gene Symbol:
          Coprs
          Gene Name:
          coordinator of PRMT5, differentiation stimulator
          Gene Aliases:
          1700029I03Rik, 2410022L05Rik, AA409325, AI256813, C85432, Copr5
          Chromosome Location:
          Chr.8: 13884787 - 13890327 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481570_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Coprs
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025556.3 NP_079832.1 3 - 4 387 67
          GenBank mRNA AK008882.1 - 3 - 4 388 67
          AK010573.1 - 3 - 4 389 67
          BC029192.1 - 3 - 4 384 2647299 67
          BC051120.1 - 1 - 2 1548 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001745.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Coprs

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          muscle organ development
          histone H4-R3 methylation
          protein binding
          histone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25671028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25671030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25671026 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25671027 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 BC051120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 NM_025556.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 AK008882.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 AK010573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 BC029192.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 BC051120.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 NM_025556.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 AK008882.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 AK010573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P357422 BC029192.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903558 AK087497.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903558 AK154085.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903558 BC026412.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903558 BC046594.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 206106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 237998 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 159548 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108972 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 BC051120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 NM_025556.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 AK008882.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 AK010573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 BC029192.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 BC051120.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 NM_025556.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 AK008882.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 AK010573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P357422 BC029192.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25671025 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01679382 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108972 - -
          Whole Mouse Genome G4122A A_51_P357422 BC051120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357422 NM_025556.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357422 AK008882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357422 AK010573.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357422 BC029192.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357422 BC051120.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357422 NM_025556.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357422 AK008882.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357422 AK010573.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357422 BC029192.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107274 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108972 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07245094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07245097 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25671026 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25671028 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25671030 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25671027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25670944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25671027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07372050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07245094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07354060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07101976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07059304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25671007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25670993 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577225 - -
          Affymetrix HT MG-430 PM Array Plate 1420113_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1448971_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1420114_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10577222 - -
          Mouse Genome 430 2.0 Array 1448971_PM_at - -
          Mouse Genome 430 2.0 Array 1448971_at - -
          Mouse Genome 430 2.0 Array 1420114_s_at - -
          Mouse Genome 430 2.0 Array 1420114_PM_s_at - -
          Mouse Genome 430 2.0 Array 1420113_PM_s_at - -
          Mouse Genome 430 2.0 Array 1420113_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66423_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66423_at - -
          Affymetrix Mouse Clariom S Array PSR0800014130.mm.1 TC0800001745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014130.mm.1 NM_025556.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014130.mm.1 NM_025556.3 Refseq
          Mouse Expression Set 430 1448971_PM_at - -
          Mouse Expression Set 430 1448971_at - -
          Murine Genome U74v2 Set 161906_PM_f_at - -
          Murine Genome U74v2 Set 96779_f_at - -
          Murine Genome U74v2 Set 96780_at - -
          Murine Genome U74v2 Set 161906_f_at - -
          Murine Genome U74v2 Set 96779_PM_f_at - -
          Murine Genome U74v2 Set 96780_PM_at - -
          Murine Genome U74v2 Set 168894_PM_at - -
          Murine Genome U74v2 Set 169889_at - -
          Murine Genome U74v2 Set 169889_PM_at - -
          Murine Genome U74v2 Set 168894_at - -
          Mouse Genome 430A 2.0 Array 1448971_PM_at - -
          Mouse Genome 430A 2.0 Array 1448971_at - -
          Affymetrix HT Mouse Genome 430A Array 1420114_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1420113_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1420114_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1448971_at - -
          Affymetrix HT Mouse Genome 430A Array 1420113_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1448971_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 878953 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 780195 - -
          Illumina mouse-6 v1.0 expression beadchip 780195 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780195 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234516 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1234516 - -
          mouse-6 v1.1 (Illumina) scl35070.6.62_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 780195 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35070.6.62_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780195 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234516 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234516 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35070.6.62_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix