Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482407_g1
          See other HIGD2A GE Assays ›
          Gene Symbol:
          Higd2a
          Gene Name:
          HIG1 domain family, member 2A
          Gene Aliases:
          2010110M21Rik, AA690237
          Chromosome Location:
          Chr.13: 54590231 - 54591147 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482407_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Higd2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025933.3 NP_080209.1 1 - 2 190 79
          GenBank mRNA AK005269.1 - 1 - 2 191 79
          AK008387.1 - 1 - 2 187 79
          BC021471.1 - 1 - 2 187 5343356 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000633.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Higd2a

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          negative regulation of apoptotic process
          mitochondrial respirasome assembly
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 AK005269.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 AK008387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 AK004272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 BC021471.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 NM_025933.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 AK005269.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 AK008387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 AK004272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 BC021471.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P450021 NM_025933.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143845 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 AK005269.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 AK008387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 AK004272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 BC021471.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 NM_025933.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 AK005269.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 AK008387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 AK004272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 BC021471.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P450021 NM_025933.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050270 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143845 - -
          Whole Mouse Genome G4122A A_51_P450021 AK005269.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450021 AK008387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450021 AK004272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450021 BC021471.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450021 NM_025933.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450021 AK005269.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450021 AK008387.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450021 AK004272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450021 BC021471.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P450021 NM_025933.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131422 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143845 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10854347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10854338 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10854340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11110029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050264 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10405289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10405290 - -
          Affymetrix HT MG-430 PM Array Plate 1418002_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10405287 - -
          Mouse Genome 430 2.0 Array 1418002_PM_at - -
          Mouse Genome 430 2.0 Array 1418002_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67044_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67044_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300004031.mm.1 TC1300000633.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004031.mm.1 NM_025933.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004031.mm.1 NM_025933.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004032.mm.1 TC1300000633.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004032.mm.1 NM_025933.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004032.mm.1 NM_025933.3 Refseq
          Mouse Expression Set 430 1418002_at - -
          Mouse Expression Set 430 1418002_PM_at - -
          Murine Genome U74v2 Set 95105_PM_at - -
          Murine Genome U74v2 Set 95105_at - -
          Mouse Genome 430A 2.0 Array 1418002_PM_at - -
          Mouse Genome 430A 2.0 Array 1418002_at - -
          Affymetrix HT Mouse Genome 430A Array 1418002_at - -
          Affymetrix HT Mouse Genome 430A Array 1418002_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 614249 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6040670 - -
          Illumina mouse-6 v1.0 expression beadchip 6040670 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006040670 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255823 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255823 - -
          mouse-6 v1.1 (Illumina) scl44752.2.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 6040670 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44752.2.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6040670 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255823 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255823 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl44752.2.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0