Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482976_g1
          See other CAPN1 GE Assays ›
          Gene Symbol:
          Capn1
          Gene Name:
          calpain 1
          Gene Aliases:
          Capa-1, Capa1, mu-calpin
          Chromosome Location:
          Chr.19: 5988545 - 6015825 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482976_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Capn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110504.1 NP_001103974.1 20 - 21 2151 94
          NM_007600.3 NP_031626.1 20 - 21 2203 94
          GenBank mRNA AF021847.1 - 20 - 21 2082 94
          AK088547.1 - 20 - 21 2123 94
          AK149090.1 - 16 - 17 1551 94
          AK149706.1 - 20 - 21 2144 94
          AK150084.1 - 20 - 21 2142 94
          AK160940.1 - 20 - 21 2143 94
          AK165258.1 - 20 - 21 2163 94
          AK168063.1 - 20 - 21 2142 94
          AK171405.1 - 20 - 21 2106 94
          BC026138.1 - 20 - 21 2118 4218148 94
          BC050276.1 - 20 - 21 2119 4923347 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000995.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Capn1

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein autoprocessing
          receptor catabolic process
          mammary gland involution
          calcium-dependent cysteine-type endopeptidase activity
          calcium ion binding
          protein binding
          cytoskeletal protein binding
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900009107.mm.1 TC1900000995.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009107.mm.1 NM_001110504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009107.mm.1 NM_001110504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009107.mm.1 NM_007600.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009107.mm.1 NM_007600.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009105.mm.1 TC1900000995.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009105.mm.1 NM_001110504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009105.mm.1 NM_001110504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009105.mm.1 NM_007600.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009105.mm.1 NM_007600.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009106.mm.1 TC1900000995.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009106.mm.1 NM_001110504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009106.mm.1 NM_001110504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009106.mm.1 NM_007600.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009106.mm.1 NM_007600.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline