Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600006624.mm.1 |
TC1600000763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006624.mm.1 |
NM_007757.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006624.mm.1 |
NM_007757.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006623.mm.1 |
TC1600000763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006623.mm.1 |
NM_007757.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006623.mm.1 |
NM_007757.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006625.mm.1 |
TC1600000763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006625.mm.1 |
NM_007757.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006625.mm.1 |
NM_007757.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006626.mm.1 |
TC1600000763.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600006626.mm.1 |
NM_007757.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600006626.mm.1 |
NM_007757.2 |
Refseq |