Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00485863_g1
          See other RRAS GE Assays ›
          Gene Symbol:
          Rras
          Gene Name:
          Harvey rat sarcoma oncogene, subgroup R
          Gene Aliases:
          AI573426
          Chromosome Location:
          Chr.7: 45018007 - 45021647 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00485863_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rras
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009101.2 NP_033127.1 5 - 6 582 83
          GenBank mRNA AK155855.1 - 5 - 6 612 83
          BC009105.1 - 5 - 6 599 3483313 83
          BC110562.1 - 5 - 6 584 40049793 83
          BC110563.1 - 5 - 6 584 40049794 83
          BC145932.1 - 5 - 6 628 40131235 83
          BC145934.1 - 5 - 6 628 40131237 83
          M21019.1 - 5 - 6 584 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000826.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rras

          Gene Ontology Categories:

          Function(s) Process(es)

          leukocyte differentiation
          signal transduction
          small GTPase mediated signal transduction
          metabolic process
          negative regulation of cell migration
          positive regulation of angiogenesis
          regulation of protein kinase B signaling
          face morphogenesis
          regulation of ERK1 and ERK2 cascade
          nucleotide binding
          GTPase activity
          GTP binding
          GDP binding
          protein-containing complex binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 BC145932.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 BC145934.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 M21019.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 AK155855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 BC009105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 BC145932.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 BC145934.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 M21019.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 AK155855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P635182 BC009105.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 888529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730312 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 791965 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160159 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 BC145932.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 BC145934.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 M21019.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 AK155855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 BC009105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 BC145932.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 BC145934.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 M21019.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 AK155855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P635182 BC009105.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 BC145932.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 BC145934.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 M21019.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 AK155855.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 BC009105.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 BC145932.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 BC145934.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 M21019.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 AK155855.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P635182 BC009105.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25090707 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160159 - -
          Whole Mouse Genome G4122A A_52_P635182 BC145932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635182 BC145934.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635182 M21019.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635182 AK155855.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635182 BC009105.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635182 BC145932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635182 BC145934.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635182 M21019.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635182 AK155855.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P635182 BC009105.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138873 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160159 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06316397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25090637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06316394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06051860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06400620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090692 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06212932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090709 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25090715 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10552829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552830 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552827 - -
          Affymetrix Mouse Gene 1.0 ST Array 10552826 - -
          Affymetrix HT MG-430 PM Array Plate 1418448_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10552824 - -
          Mouse Genome 430 2.0 Array 1418448_at - -
          Mouse Genome 430 2.0 Array 1418448_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20130_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20130_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700007050.mm.1 TC0700000826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700007050.mm.1 NM_009101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007050.mm.1 NM_009101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007048.mm.1 TC0700000826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700007048.mm.1 NM_009101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007048.mm.1 NM_009101.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007049.mm.1 TC0700000826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700007049.mm.1 NM_009101.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700007049.mm.1 NM_009101.2 Refseq
          Mouse Expression Set 430 1418448_PM_at - -
          Mouse Expression Set 430 1418448_at - -
          Murine Genome U74v2 Set 94394_at - -
          Murine Genome U74v2 Set 94394_PM_at - -
          Mouse Genome 430A 2.0 Array 1418448_PM_at - -
          Mouse Genome 430A 2.0 Array 1418448_at - -
          Affymetrix HT Mouse Genome 430A Array 1418448_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418448_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 543547 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4150609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004150609 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658804 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658804 - -
          mouse-6 v1.1 (Illumina) scl32699.8.1_168-S - -
          Illumina Sentrix mouse-6.v1 bead array 4150609 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32699.8.1_168-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4150609 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2658804 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2658804 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32699.8.1_168-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0