Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300009799.mm.1 |
TC0300001198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009799.mm.1 |
NM_001113478.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009799.mm.1 |
NM_001113478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009799.mm.1 |
NM_009146.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009799.mm.1 |
NM_009146.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009804.mm.1 |
TC0300001198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009804.mm.1 |
NM_001113478.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009804.mm.1 |
NM_001113478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009804.mm.1 |
NM_009146.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009804.mm.1 |
NM_009146.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009800.mm.1 |
TC0300001198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009800.mm.1 |
NM_001113478.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009800.mm.1 |
NM_001113478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009800.mm.1 |
NM_009146.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009800.mm.1 |
NM_009146.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009803.mm.1 |
TC0300001198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009803.mm.1 |
NM_001113478.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009803.mm.1 |
NM_001113478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009803.mm.1 |
NM_009146.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009803.mm.1 |
NM_009146.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009805.mm.1 |
TC0300001198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009805.mm.1 |
NM_001113478.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009805.mm.1 |
NM_001113478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009805.mm.1 |
NM_009146.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009805.mm.1 |
NM_009146.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009798.mm.1 |
TC0300001198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009798.mm.1 |
NM_001113478.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009798.mm.1 |
NM_001113478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009798.mm.1 |
NM_009146.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009798.mm.1 |
NM_009146.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009801.mm.1 |
TC0300001198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009801.mm.1 |
NM_001113478.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009801.mm.1 |
NM_001113478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009801.mm.1 |
NM_009146.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009801.mm.1 |
NM_009146.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009802.mm.1 |
TC0300001198.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300009802.mm.1 |
NM_001113478.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009802.mm.1 |
NM_001113478.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009802.mm.1 |
NM_009146.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300009802.mm.1 |
NM_009146.3 |
Refseq |