Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900006433.mm.1 |
TC1900000723.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006433.mm.1 |
NM_001164639.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006433.mm.1 |
NM_001164639.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006433.mm.1 |
NM_009289.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006433.mm.1 |
NM_009289.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006434.mm.1 |
TC1900000723.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006434.mm.1 |
NM_001164639.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006434.mm.1 |
NM_001164639.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006434.mm.1 |
NM_009289.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006434.mm.1 |
NM_009289.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006432.mm.1 |
TC1900000723.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006432.mm.1 |
NM_001164639.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006432.mm.1 |
NM_001164639.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006432.mm.1 |
NM_009289.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006432.mm.1 |
NM_009289.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006435.mm.1 |
TC1900000723.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006435.mm.1 |
NM_001164639.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006435.mm.1 |
NM_001164639.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006435.mm.1 |
NM_009289.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006435.mm.1 |
NM_009289.3 |
Refseq |