Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00486756_m1
          See other CTNNA1 GE Assays ›
          Gene Symbol:
          Ctnna1
          Gene Name:
          catenin (cadherin associated protein), alpha 1
          Gene Aliases:
          2010010M04Rik, AA517462, AI988031, Catna1
          Chromosome Location:
          Chr.18: 35118912 - 35254775 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00486756_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctnna1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009818.1 NP_033948.1 14 - 15 2083 96
          GenBank mRNA AK004623.1 - 14 - 15 2041 96
          AK036752.1 - 14 - 15 2108 96
          AK046513.1 - 8 - 9 1442 96
          AK148496.1 - 14 - 15 2080 96
          AK149721.1 - 14 - 15 2098 96
          AK149738.1 - 14 - 15 2098 96
          AK159339.1 - 14 - 15 2098 96
          AK168635.1 - 14 - 15 2108 96
          BC048163.1 - 14 - 15 2071 6418358 96
          BC060174.1 - 14 - 15 2078 96
          BC068316.1 - 14 - 15 2075 30106659 96
          BC105576.1 - 14 - 15 2063 3967124 96
          D90362.1 - 14 - 15 2074 96
          X59990.1 - 14 - 15 2083 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000306.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctnna1

          Gene Ontology Categories:

          Function(s) Process(es)

          actin filament organization
          cell adhesion
          establishment or maintenance of cell polarity
          negative regulation of neuroblast proliferation
          gap junction assembly
          cellular protein localization
          regulation of cell population proliferation
          negative regulation of apoptotic process
          apical junction assembly
          positive regulation of smoothened signaling pathway
          protein heterooligomerization
          cellular response to indole-3-methanol
          epithelial cell-cell adhesion
          negative regulation of cell motility
          negative regulation of integrin-mediated signaling pathway
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          RNA binding
          structural molecule activity
          protein binding
          beta-catenin binding
          vinculin binding
          gamma-catenin binding
          cadherin binding
          actin filament binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800002251.mm.1 TC1800000306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002251.mm.1 NM_009818.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002251.mm.1 NM_009818.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002252.mm.1 TC1800000306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002252.mm.1 NM_009818.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002252.mm.1 NM_009818.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002250.mm.1 TC1800000306.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002250.mm.1 NM_009818.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002250.mm.1 NM_009818.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0