Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00489036_m1
          See other SP1 GE Assays ›
          Gene Symbol:
          Sp1
          Gene Name:
          trans-acting transcription factor 1
          Gene Aliases:
          1110003E12Rik, AA450830, AI845540, Sp1-1
          Chromosome Location:
          Chr.15: 102406316 - 102436404 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00489036_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013672.2 NP_038700.2 1 - 2 109 121
          GenBank mRNA AF022363.1 - 1 - 2 97 121
          CJ181598.1 - 1 - 2 116 121
          EF200005.1 - 1 - 2 113 121
          S79832.1 - 1 - 2 100 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001098.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AF022363.1, EF200005.1, NM_013672.2, S79832.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sp1

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          in utero embryonic development
          trophectodermal cell differentiation
          liver development
          embryonic placenta development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          megakaryocyte differentiation
          lung development
          enucleate erythrocyte differentiation
          positive regulation by host of viral transcription
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          rhythmic process
          embryonic camera-type eye morphogenesis
          embryonic skeletal system development
          embryonic process involved in female pregnancy
          definitive hemopoiesis
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          cis-regulatory region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          nucleic acid binding
          DNA binding
          double-stranded DNA binding
          DNA-binding transcription factor activity
          protein binding
          protein C-terminus binding
          transcription factor binding
          histone acetyltransferase binding
          protein homodimerization activity
          histone deacetylase binding
          bHLH transcription factor binding
          sequence-specific DNA binding
          metal ion binding
          RNA polymerase II-specific DNA-binding transcription factor binding
          HMG box domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500008815.mm.1 TC1500001098.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008815.mm.1 NM_013672.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008815.mm.1 NM_013672.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008809.mm.1 TC1500001098.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008809.mm.1 NM_013672.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008809.mm.1 NM_013672.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008808.mm.1 TC1500001098.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500008808.mm.1 NM_013672.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500008808.mm.1 NM_013672.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0