Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500003868.mm.1 |
TC1500000561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003868.mm.1 |
NM_001163465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003868.mm.1 |
NM_001163465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003868.mm.1 |
NM_008164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003868.mm.1 |
NM_008164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003869.mm.1 |
TC1500000561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003869.mm.1 |
NM_001163465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003869.mm.1 |
NM_001163465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003869.mm.1 |
NM_008164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003869.mm.1 |
NM_008164.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003866.mm.1 |
TC1500000561.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500003866.mm.1 |
NM_001163465.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003866.mm.1 |
NM_001163465.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003866.mm.1 |
NM_008164.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003866.mm.1 |
NM_008164.2 |
Refseq |