Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008513.3 | NP_032539.2 | 2 - 3 | 601 | 87 | |
GenBank mRNA | AF064984.1 | - | 2 - 3 | 591 | 87 | |
AF077847.1 | - | 2 - 3 | 479 | 87 | ||
AK147068.1 | - | 2 - 3 | 594 | 87 | ||
AK147231.1 | - | 2 - 3 | 599 | 87 | ||
AK147330.1 | - | 2 - 3 | 599 | 87 | ||
AK220186.1 | - | 2 - 3 | 554 | 87 | ||
BC011374.1 | - | 2 - 3 | 583 | 3154246 | 87 | |
BC094610.1 | - | 2 - 3 | 590 | 87 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |