Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00493611_g1
          See other TEX261 GE Assays ›
          Gene Symbol:
          Tex261
          Gene Name:
          testis expressed gene 261
          Gene Aliases:
          3110001O07Rik, AA409339, AI480706, AL033351, TEG-261
          Chromosome Location:
          Chr.6: 83768881 - 83775814 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00493611_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex261
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009357.2 NP_033383.1 5 - 6 560 65
          GenBank mRNA AK149789.1 - 5 - 6 525 65
          BC003802.1 - 5 - 6 504 3486968 65
          X81058.1 - 5 - 6 506 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002582.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tex261

          Gene Ontology Categories:

          Function(s) Process(es)

          endoplasmic reticulum to Golgi vesicle-mediated transport
          positive regulation of apoptotic process
          molecular_function
          COPII receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595446 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595444 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595442 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242733 BC003802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242733 X80425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242733 X81058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242733 AK157834.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242733 AK170049.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 608253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 293923 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 657829 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14510 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121320 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242733 BC003802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242733 X80425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242733 X81058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242733 AK157834.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242733 AK170049.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595465 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595472 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595468 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595462 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595460 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595439 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595473 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24595435 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121320 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05962527 - -
          Whole Mouse Genome G4122A A_51_P147654 AK170049.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P147654 AK013969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P147654 AK013971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P378452 AK013969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P378452 AK013971.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P654021 BC003802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P654021 X81058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P654021 AK149789.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P654021 NM_009357.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129436 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121320 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05256487 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05298582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05298584 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05292001 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595444 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595446 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24595445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24595458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05256486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05298601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05425888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05108999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05298600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05730233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05298602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05298568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05298572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05032321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05818596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05730234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595445 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24595398 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545757 - -
          Affymetrix HT MG-430 PM Array Plate 1450965_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423384_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1452768_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545751 - -
          Mouse Genome 430 2.0 Array 1423384_s_at - -
          Mouse Genome 430 2.0 Array 1450965_at - -
          Mouse Genome 430 2.0 Array 1452768_at - -
          Mouse Genome 430 2.0 Array 1452768_PM_at - -
          Mouse Genome 430 2.0 Array 1450965_PM_at - -
          Mouse Genome 430 2.0 Array 1423384_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21766_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21766_at - -
          Affymetrix Mouse Clariom S Array PSR0600020084.mm.1 TC0600002582.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020084.mm.1 NM_009357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020084.mm.1 NM_009357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020085.mm.1 TC0600002582.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020085.mm.1 NM_009357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020085.mm.1 NM_009357.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020086.mm.1 TC0600002582.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020086.mm.1 NM_009357.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020086.mm.1 NM_009357.2 Refseq
          Mouse Expression Set 430 1423384_s_at - -
          Mouse Expression Set 430 1450965_at - -
          Mouse Expression Set 430 1423384_PM_s_at - -
          Mouse Expression Set 430 1450965_PM_at - -
          Murine Genome U74v2 Set 96166_PM_at - -
          Murine Genome U74v2 Set 101517_at - -
          Murine Genome U74v2 Set 96166_at - -
          Murine Genome U74v2 Set 101517_PM_at - -
          Mouse Genome 430A 2.0 Array 1423384_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1423384_s_at - -
          Mouse Genome 430A 2.0 Array 1450965_at - -
          Mouse Genome 430A 2.0 Array 1450965_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450965_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423384_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452768_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423384_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1450965_at - -
          Affymetrix HT Mouse Genome 430A Array 1452768_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2340037 - -
          Illumina mouse-6 v1.0 expression beadchip 2630672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002630672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253322 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605494 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253322 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605494 - -
          mouse-6 v1.1 (Illumina) scl28784.6_160-S - -
          mouse-6 v1.1 (Illumina) scl0021766.1_289-S - -
          Illumina Sentrix mouse-6.v1 bead array 2630672 - -
          Illumina Sentrix mouse-6.v1 bead array 2340037 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0021766.1_289-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28784.6_160-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340037 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2630672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605494 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1253322 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253322 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605494 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0021766.1_289-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl28784.6_160-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0