Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00494622_s1
          See other GYKL1 GE Assays ›
          Gene Symbol:
          Gykl1
          Gene Name:
          glycerol kinase-like 1
          Gene Aliases:
          AI449806, Gk-rs1
          Chromosome Location:
          Chr.18: 52693679 - 52695593 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00494622_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gykl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010293.3 NP_034423.2 1 1455 63
          GenBank mRNA AF117733.1 - 1 1455 63
          AK076631.1 - 1 1431 63
          BC050968.1 - 1 1444 63
          BC119574.2 - 1 1436 40058020 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000527.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gykl1

          Gene Ontology Categories:

          Function(s) Process(es)

          glycerol metabolic process
          triglyceride metabolic process
          phosphorylation
          glycerol-3-phosphate biosynthetic process
          glycerol kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P389396 AK076631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P389396 BC050968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P389396 AF117733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P389396 NM_010293.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P389396 AK076631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P389396 BC050968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P389396 AF117733.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P389396 NM_010293.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658131 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170242 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179056 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167336 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P389396 AK076631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P389396 BC050968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P389396 AF117733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P389396 NM_010293.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P389396 AK076631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P389396 BC050968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P389396 AF117733.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P389396 NM_010293.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P389396 AK076631.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P389396 BC050968.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P389396 AF117733.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P389396 NM_010293.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P389396 AK076631.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P389396 BC050968.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P389396 AF117733.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P389396 NM_010293.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31708998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31708997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31709047 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170242 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167336 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179056 - -
          Whole Mouse Genome G4122A A_51_P389396 AK076631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P389396 BC050968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P389396 AF117733.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P389396 NM_010293.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P389396 AK076631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P389396 BC050968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P389396 AF117733.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P389396 NM_010293.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135370 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167336 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179056 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170242 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13819841 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13819839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31708983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31709010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31708997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31708990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13929267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13624692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13819827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31708994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31708983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709042 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13624694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31708995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31708986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709075 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13952904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31708993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13624695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13716385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13616243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31708996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31709057 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455720 - -
          Affymetrix HT MG-430 PM Array Plate 1423514_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455719 - -
          Mouse Genome 430 2.0 Array 1423514_at - -
          Mouse Genome 430 2.0 Array 1423514_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14625_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14625_at - -
          Affymetrix Mouse Clariom S Array PSR1800003816.mm.1 TC1800000527.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800003816.mm.1 NM_010293.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800003816.mm.1 NM_010293.3 Refseq
          Affymetrix HT Mouse Genome 430A Array 1423514_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423514_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 865593 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6900121 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006900121 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2665209 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2665209 - -
          mouse-6 v1.1 (Illumina) scl51953.1.12_171-S - -
          Illumina Sentrix mouse-6.v1 bead array 6900121 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51953.1.12_171-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6900121 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2665209 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2665209 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0