Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00497250_m1
          See other LSM4 GE Assays ›
          Gene Symbol:
          Lsm4
          Gene Name:
          LSM4 homolog, U6 small nuclear RNA associated (S. cerevisiae)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 70673231 - 70678752 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00497250_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lsm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015816.4 NP_056631.2 1 - 2 228 86
          GenBank mRNA AJ249439.1 - 1 - 2 68 86
          AK003847.1 - 1 - 2 64 86
          AK010910.1 - 1 - 2 64 86
          AK089028.1 - 1 - 2 81 86
          AK143470.1 - 1 - 2 52 86
          AK167784.1 - 1 - 2 86
          BC026747.1 - 1 - 2 211 4024321 86
          CA322716.1 - 1 - 2 228 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000759.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AJ249439.1, AK003847.1, AK010910.1, AK089028.1, AK143470.1, AK167784.1, BC026747.1, NM_015816.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lsm4

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          mRNA processing
          RNA splicing
          positive regulation of mitophagy in response to mitochondrial depolarization
          RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953305 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953309 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953303 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953308 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953307 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953304 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P116487 AJ249439.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P116487 AJ249439.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356749 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132899 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P116487 AJ249439.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P116487 AJ249439.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P116487 AJ249439.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P116487 AJ249439.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25953322 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132899 - -
          Whole Mouse Genome G4122A A_51_P116487 AJ249439.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P116487 AJ249439.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131252 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132899 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07227532 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07227534 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07111224 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07111223 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953305 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953303 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953307 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953309 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953304 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25953308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25953342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06816557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06725002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07253418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07231665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07027777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07167907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07111225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07073514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06816552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07227534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07122948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06816551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06914049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07253411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06813408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06940649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06824196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07030842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06914051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25953342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06856563 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10572455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572451 - -
          Affymetrix HT MG-430 PM Array Plate 1448622_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10572449 - -
          Mouse Genome 430 2.0 Array 1448622_PM_at - -
          Mouse Genome 430 2.0 Array 1448622_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50783_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50783_at - -
          Affymetrix Mouse Clariom S Array PSR0800005378.mm.1 TC0800000759.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005378.mm.1 NM_015816.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005378.mm.1 NM_015816.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005380.mm.1 TC0800000759.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005380.mm.1 NM_015816.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005380.mm.1 NM_015816.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005379.mm.1 TC0800000759.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005379.mm.1 NM_015816.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005379.mm.1 NM_015816.4 Refseq
          Mouse Expression Set 430 1448622_PM_at - -
          Mouse Expression Set 430 1448622_at - -
          Murine Genome U74v2 Set 93008_at - -
          Murine Genome U74v2 Set 93008_PM_at - -
          Mouse Genome 430A 2.0 Array 1448622_PM_at - -
          Mouse Genome 430A 2.0 Array 1448622_at - -
          Affymetrix HT Mouse Genome 430A Array 1448622_at - -
          Affymetrix HT Mouse Genome 430A Array 1448622_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 916748 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3130180 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130180 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2763960 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2763960 - -
          mouse-6 v1.1 (Illumina) scl33713.6.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 3130180 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33713.6.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2903511 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2903511 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33713.6.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0