Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00497908_m1
          See other LGALS12 GE Assays ›
          Gene Symbol:
          Lgals12
          Gene Name:
          lectin, galactose binding, soluble 12
          Gene Aliases:
          AW987437, Grip1
          Chromosome Location:
          Chr.19: 7596656 - 7610466 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00497908_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lgals12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019516.3 NP_062389.1 7 - 8 1099 64
          GenBank mRNA AF223223.1 - 8 - 9 793 64
          AF244978.1 - 8 - 9 801 64
          AF244979.1 - 7 - 8 846 64
          AK033535.1 - 6 - 7 987 64
          AK085660.1 - 7 - 8 1102 64
          AK136526.1 - 7 - 8 1100 64
          BC030890.1 - 7 - 8 783 4485119 64

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lgals12

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          intrinsic apoptotic signaling pathway
          carbohydrate binding
          lactose binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31940698 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31940699 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31940700 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092204 BC030890.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092204 AK085660.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092204 AK136526.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092204 AF244979.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092204 AK033535.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 951910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228998 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123471 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144102 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092204 BC030890.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092204 AK085660.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092204 AK136526.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092204 AF244979.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092204 AK033535.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P293482 BC030890.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P293482 AK085660.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P293482 AK136526.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P293482 AK033535.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P293482 AK079940.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31940683 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123471 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03546542 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144102 - -
          Whole Mouse Genome G4122A A_52_P293482 BC030890.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P293482 AK085660.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P293482 AK136526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P293482 AK033535.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P293482 AK079940.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113324 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123471 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144102 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14359817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14230433 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14359814 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14492556 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14239955 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14359827 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14470948 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14223579 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31940698 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31940699 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31940700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31940732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14360908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14360905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14323462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14230433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14511657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14227737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14223579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14170504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31940690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14360920 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465709 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465704 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465707 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465710 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465703 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465702 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465708 - -
          Affymetrix HT MG-430 PM Array Plate 1417686_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417687_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465701 - -
          Mouse Genome 430 2.0 Array 1417686_at - -
          Mouse Genome 430 2.0 Array 1417686_PM_at - -
          Mouse Genome 430 2.0 Array 1417687_at - -
          Mouse Genome 430 2.0 Array 1417687_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56072_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56072_PM_at - -
          Mouse Expression Set 430 1417687_at - -
          Mouse Expression Set 430 1417687_PM_at - -
          Mouse Expression Set 430 1417686_PM_at - -
          Mouse Expression Set 430 1417686_at - -
          Murine Genome U74v2 Set 166408_PM_r_at - -
          Murine Genome U74v2 Set 166408_r_at - -
          Mouse Genome 430A 2.0 Array 1417686_at - -
          Mouse Genome 430A 2.0 Array 1417687_at - -
          Mouse Genome 430A 2.0 Array 1417686_PM_at - -
          Mouse Genome 430A 2.0 Array 1417687_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417686_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417686_at - -
          Affymetrix HT Mouse Genome 430A Array 1417687_at - -
          Affymetrix HT Mouse Genome 430A Array 1417687_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3940242 - -
          Illumina mouse-6 v1.0 expression beadchip 3940242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2761326 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2761326 - -
          mouse-6 v1.1 (Illumina) scl056072.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056072.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2761326 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2761326 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0