Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499163_m1
          See other RALY GE Assays ›
          Gene Symbol:
          Raly
          Gene Name:
          hnRNP-associated with lethal yellow
          Gene Aliases:
          AI663842, Merc
          Chromosome Location:
          Chr.2: 154791097 - 154871010 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499163_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Raly
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001139511.1 NP_001132983.1 2 - 3 705 96
          NM_001139512.1 NP_001132984.1 3 - 4 471 96
          NM_001139513.1 NP_001132985.1 2 - 3 705 96
          NM_023130.3 NP_075619.2 3 - 4 788 96
          GenBank mRNA AK014356.1 - 3 - 4 541 96
          AK135386.1 - 2 - 3 464 96
          AK152290.1 - 3 - 4 547 96
          AK154787.1 - 2 - 3 462 96
          AK168781.1 - 3 - 4 545 96
          BC004851.1 - 2 - 3 449 2810841 96
          BC016587.1 - 3 - 4 537 2811315 96
          CX214615.1 - 2 - 3 448 96
          L17076.1 - 3 - 4 518 96
          S72641.1 - 3 - 4 739 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002352.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Raly

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          nucleotide binding
          nucleic acid binding
          RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200018711.mm.1 TC0200002352.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200018711.mm.1 NM_001139511.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018711.mm.1 NM_001139511.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018711.mm.1 NM_001139512.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018711.mm.1 NM_001139512.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018711.mm.1 NM_001139513.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018711.mm.1 NM_001139513.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018711.mm.1 NM_023130.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018711.mm.1 NM_023130.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018710.mm.1 TC0200002352.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200018710.mm.1 NM_001139511.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018710.mm.1 NM_001139511.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018710.mm.1 NM_001139512.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018710.mm.1 NM_001139512.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018710.mm.1 NM_001139513.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018710.mm.1 NM_001139513.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018710.mm.1 NM_023130.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200018710.mm.1 NM_023130.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0