Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00502981_m1
          See other TMEM54 GE Assays ›
          Gene Symbol:
          Tmem54
          Gene Name:
          transmembrane protein 54
          Gene Aliases:
          1810017F10Rik, AI507302
          Chromosome Location:
          Chr.4: 129105515 - 129111626 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00502981_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem54
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290706.1 NP_001277635.1 1 - 2 186 59
          NM_025452.4 NP_079728.2 1 - 2 204 59
          NM_133881.1 NP_598642.1 1 - 2 204 59
          GenBank mRNA AK007525.1 - 1 - 2 119 59
          AK008935.1 - 1 - 2 119 59
          AK009830.1 - 1 - 2 100 59
          BC019563.1 - 1 - 2 133 4219997 59
          BY079646.1 - 1 - 2 152 59
          BY092384.1 - 1 - 2 186 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001512.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK008935.1, AK009830.1, BC019563.1, NM_001290706.1, NM_025452.4, NM_133881.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem54

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23294937 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23294939 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23294938 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23294940 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181888 BQ032787.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181888 NM_025452.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181888 NM_133881.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181888 AK008935.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2181888 AK009830.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63692 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113888 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181888 BQ032787.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181888 NM_025452.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181888 NM_133881.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181888 AK008935.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2181888 AK009830.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23294921 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113888 - -
          Whole Mouse Genome G4122A A_51_P303079 BQ032787.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303079 AK007525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303079 NM_025452.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303079 NM_133881.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P303079 AK008935.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129867 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140301 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03730173 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03730179 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03576238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23294938 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23294937 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23294939 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23294940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23295017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23294934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03695982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03486004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04101060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03943501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04101049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03486005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03486006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03360161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03695985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03968557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04101057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03231354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03322268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03231349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04101059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04101056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03943500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03231355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03850091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03855096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03632615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03855098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03943502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04101058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23294924 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10508411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10508407 - -
          Affymetrix HT MG-430 PM Array Plate 1417895_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10508405 - -
          Mouse Genome 430 2.0 Array 1417895_PM_a_at - -
          Mouse Genome 430 2.0 Array 1417895_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66260_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66260_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400011638.mm.1 TC0400001512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011638.mm.1 NM_025452.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011638.mm.1 NM_025452.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011638.mm.1 NM_133881.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011638.mm.1 NM_133881.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011640.mm.1 TC0400001512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011640.mm.1 NM_025452.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011640.mm.1 NM_025452.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011640.mm.1 NM_133881.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011640.mm.1 NM_133881.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011642.mm.1 TC0400001512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011642.mm.1 NM_025452.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011642.mm.1 NM_025452.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011642.mm.1 NM_133881.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011642.mm.1 NM_133881.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011639.mm.1 TC0400001512.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011639.mm.1 NM_025452.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011639.mm.1 NM_025452.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011639.mm.1 NM_133881.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011639.mm.1 NM_133881.1 Refseq
          Affymetrix HT Mouse Genome 430A Array 1417895_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417895_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 906092 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 780059 - -
          Illumina mouse-6 v1.0 expression beadchip 780059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780059 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241297 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1241297 - -
          mouse-6 v1.1 (Illumina) scl24923.6.1_23-S - -
          Illumina Sentrix mouse-6.v1 bead array 780059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24923.6.1_23-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780059 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2919393 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2919393 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline