Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133895.1 | NP_598656.1 | 1 - 2 | 582 | 110 | |
GenBank mRNA | AK154238.1 | - | 1 - 2 | 609 | 110 | |
AK182965.1 | - | 1 - 2 | 75 | 110 | ||
AK199205.1 | - | 1 - 2 | 75 | 110 | ||
AK214917.1 | - | 1 - 2 | 75 | 110 | ||
AY050630.1 | - | 1 - 2 | 588 | 110 | ||
BC016233.1 | - | 1 - 2 | 582 | 4502184 | 110 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |