Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00506108_m1
          See other ZC3H10 GE Assays ›
          Gene Symbol:
          Zc3h10
          Gene Name:
          zinc finger CCCH type containing 10
          Gene Aliases:
          AI790326
          Chromosome Location:
          Chr.10: 128543563 - 128547744 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00506108_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zc3h10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_134003.1 NP_598764.1 2 - 3 119 71
          GenBank mRNA AK031312.1 - 150 71
          AK050662.1 - 2 - 3 113 71
          BC022677.1 - 2 - 3 119 4239511 71

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK050662.1, BC022677.1, NM_134003.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zc3h10

          Gene Ontology Categories:

          Process(es)

          RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541967 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541960 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541961 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329158 BC022677.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329158 AK031312.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329158 AK050662.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329158 NM_134003.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329158 BC022677.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329158 AK031312.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329158 AK050662.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P329158 NM_134003.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 737821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 25103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122387 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160653 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329158 BC022677.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329158 AK031312.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329158 AK050662.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329158 NM_134003.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329158 BC022677.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329158 AK031312.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329158 AK050662.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P329158 NM_134003.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27541950 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160653 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07233437 - -
          Whole Mouse Genome G4122A A_52_P405525 AK031312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329158 BC022677.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329158 AK031312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329158 AK050662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329158 NM_134003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329158 BC022677.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329158 AK031312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329158 AK050662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P329158 NM_134003.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131872 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160653 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08592370 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08592374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08592390 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541961 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541962 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541966 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541967 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27541960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27541975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27542007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08702732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08702731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08592390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08543151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08363540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27541955 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373444 - -
          Affymetrix HT MG-430 PM Array Plate 1417833_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373439 - -
          Mouse Genome 430 2.0 Array 1417833_PM_at - -
          Mouse Genome 430 2.0 Array 1417833_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 103284_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 103284_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000023658.mm.1 TC1000003150.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023658.mm.1 NM_134003.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023658.mm.1 NM_134003.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023654.mm.1 TC1000003150.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023654.mm.1 NM_134003.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023654.mm.1 NM_134003.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023653.mm.1 TC1000003150.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023653.mm.1 NM_134003.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023653.mm.1 NM_134003.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023655.mm.1 TC1000003150.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023655.mm.1 NM_134003.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023655.mm.1 NM_134003.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023656.mm.1 TC1000003150.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023656.mm.1 NM_134003.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023656.mm.1 NM_134003.1 Refseq
          Mouse Expression Set 430 1417833_PM_at - -
          Mouse Expression Set 430 1417833_at - -
          Murine Genome U74v2 Set 167559_i_at - -
          Murine Genome U74v2 Set 167559_PM_i_at - -
          Mouse Genome 430A 2.0 Array 1417833_at - -
          Mouse Genome 430A 2.0 Array 1417833_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417833_at - -
          Affymetrix HT Mouse Genome 430A Array 1417833_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 606974 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4010280 - -
          Illumina mouse-6 v1.0 expression beadchip 4010280 - -
          Illumina mouse-6 v1.0 expression beadchip 101090091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643115 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643115 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2469075 - -
          mouse-6 v1.1 (Illumina) scl37358.5_279-S - -
          mouse-6 v1.1 (Illumina) scl00103284.2_177-S - -
          Illumina Sentrix mouse-6.v1 bead array 101090091 - -
          Illumina Sentrix mouse-6.v1 bead array 4010280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00103284.2_177-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010280 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101090091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2643115 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643115 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2469075 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00103284.2_177-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc