Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509247_m1
          See other TMEM167B GE Assays ›
          Gene Symbol:
          Tmem167b
          Gene Name:
          transmembrane protein 167B
          Gene Aliases:
          2010200O16Rik
          Chromosome Location:
          Chr.3: 108556425 - 108562466 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509247_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem167b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026198.2 NP_080474.2 1 - 2 197 69
          GenBank mRNA AK008415.1 - 1 - 2 110 69
          AK046697.1 - 1 - 2 98 69
          BC050932.1 - 1 - 2 101 6811263 69
          BC055449.1 - 1 - 2 114 3993993 69
          BQ560740.1 - 1 - 2 197 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002746.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK008415.1, AK046697.1, BC050932.1, BC055449.1, NM_026198.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem167b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413376 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413426 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413313 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413374 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413373 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413317 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413425 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413315 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413314 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413375 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413316 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413377 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072288 BC050932.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072288 AK017252.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072288 NM_026198.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072288 AK008415.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 688296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 202101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 50114 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106820 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072288 BC050932.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072288 AK017252.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072288 NM_026198.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072288 AK008415.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413385 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22413405 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106820 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05025503 - -
          Whole Mouse Genome G4122A A_51_P348985 BC050932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P348985 AK046697.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P348985 AK155399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P348985 NM_026198.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P348985 AK008415.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101408 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106820 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02733799 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02937808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02733796 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02656881 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02937807 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413377 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413374 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413426 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413376 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413375 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413313 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413425 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413314 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413315 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413317 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413316 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22413373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22413326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03021507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02937805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02872111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02937806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03171023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03185959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03171028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02828057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02733799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02733796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02928632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02331205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02928637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03061430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03021517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03185960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03021514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02937804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03021513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02921515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02770462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22413361 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10501401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501399 - -
          Affymetrix Mouse Gene 1.0 ST Array 10501400 - -
          Affymetrix HT MG-430 PM Array Plate 1429042_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452998_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10501397 - -
          Mouse Genome 430 2.0 Array 1452998_at - -
          Mouse Genome 430 2.0 Array 1429042_at - -
          Mouse Genome 430 2.0 Array 1429042_PM_at - -
          Mouse Genome 430 2.0 Array 1452998_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67495_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67495_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300021616.mm.1 TC0300002746.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300021616.mm.1 NM_026198.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021616.mm.1 NM_026198.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021619.mm.1 TC0300002746.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300021619.mm.1 NM_026198.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021619.mm.1 NM_026198.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021621.mm.1 TC0300002746.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300021621.mm.1 NM_026198.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300021621.mm.1 NM_026198.2 Refseq
          Mouse Expression Set 430 1429042_PM_at - -
          Mouse Expression Set 430 1452998_PM_at - -
          Mouse Expression Set 430 1429042_at - -
          Mouse Expression Set 430 1452998_at - -
          Affymetrix HT Mouse Genome 430B Array 1429042_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452998_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429042_at - -
          Affymetrix HT Mouse Genome 430B Array 1452998_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 803512 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 730440 - -
          Illumina mouse-6 v1.0 expression beadchip 730440 - -
          Illumina mouse-6 v1.0 expression beadchip 100540575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730440 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215233 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215233 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236287 - -
          mouse-6 v1.1 (Illumina) scl0067495.2_241-S - -
          mouse-6 v1.1 (Illumina) scl21645.4_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 100540575 - -
          Illumina Sentrix mouse-6.v1 bead array 730440 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0067495.2_241-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730440 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100540575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215233 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215233 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0067495.2_241-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0